Skip to Content
Merck

M0187

Greiner CELLSTAR® 96 well plates

flat bottom white wells (chimney style), white polystyrene plate, lid, with condensation rings, sterile, pack of 32 ea 4 packs of 8ea

Synonym(s):

96 multiwell plates, 96 well culture plates, 96 well microplates, 96 well microtiter plates, 96 well plates, Greiner 655083

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
white polystyrene plate, flat bottom white wells (chimney style)
Size:
96 wells
Sterility:
sterile
Binding type:
Tissue Culture (TC)-treated surface
Feature:
lid with condensation rings, skirt (F-bottom)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

white polystyrene plate, flat bottom white wells (chimney style)

sterility

sterile

feature

lid with condensation rings, skirt (F-bottom)

packaging

pack of 32 ea 4 packs of 8ea

manufacturer/tradename

Greiner 655083

maximum volume

392 μL

size

96 wells

working volume

340 μL

binding type

Tissue Culture (TC)-treated surface

General description

CELLSTAR® 96W Microplate, well working volume 25-340 μL; total well volume 392 μL; growth area 0.34 cm(2).
Tissue culture treated for optimal cell attachment. Non-pyrogenic by LAL method to <0.06 EU/mL.

Application

White plates minimize cross talk in luminescence procedures. Suitable for cell work. Free of detectable DNase/RNase, human DNA and pyrogens.

Other Notes

96 well flat (F) solid bottom; cell binding microplate, with lid, Chimney Style; alphanumeric well coding, length: 127.76 mm; width: 85.48 mm; curvature approx. 200 um.

Legal Information

CELLSTAR is a registered trademark of Greiner Bio-One GmbH


Still not finding the right product?

Explore all of our products under Greiner CELLSTAR® 96 well plates




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library