Skip to Content
Merck
All Photos(3)

Key Documents

L9650

Sigma-Aldrich

Lithium chloride

for molecular biology, ≥99%

Synonym(s):

LiCl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiCl
CAS Number:
Molecular Weight:
42.39
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31
grade:
Molecular Biology
for molecular biology
form:
powder
solubility:
H2O: 100 mg/mL at 20 °C, clear, colorless

grade

Molecular Biology
for molecular biology

Assay

≥99%

form

powder

mp

605 °C (lit.)

solubility

H2O: 100 mg/mL at 20 °C, clear, colorless

anion traces

sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.003%
Fe: ≤0.001%
K: ≤0.01%
Na: ≤0.20%
heavy metals (as Pb): ≤0.002%

foreign activity

DNase, NICKase, RNase, protease, none detected

SMILES string

[Li+].[Cl-]

InChI

1S/ClH.Li/h1H;/q;+1/p-1

InChI key

KWGKDLIKAYFUFQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Lithium chloride has been used:
  • in the preparation of wash buffer for chromatin immunoprecipitation
  • in 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyl tetrazolium bromide (MTT) assay
  • to treat aged myoblast as glycogen synthase kinase (GSK) inhibitor to analyse its effect on wingless/tailess (wnt) signaling

May be used to selectively pre­cipitate RNA.

Biochem/physiol Actions

Lithium chloride has the ability to block glycogen synthase kinase (GSK). It may also possess anti-inflammatory effects at low and non-toxic concentrations.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

mTOR and neuroinflammation
Molecules to Medicine with mTOR, 317-329 (2016)
Wnt10b deficiency promotes coexpression of myogenic and adipogenic programs in myoblasts
Vertino AM, et al.
Molecular Biology of the Cell, 16(4), 2039-2048 (2005)
A laboratory practical illustrating the use of the ChIP-qPCR method in a robust model: Estrogen receptor alpha immunoprecipitation using Mcf-7 culture cells
Lacazette E
Biochemistry and Molecular Biology Education, 45(2), 152-160 (2017)
Laforin negatively regulates cell cycle progression through glycogen synthase kinase 3beta-dependent mechanisms
Liu R, et al.
Molecular and cellular biology, 28(23), 7236-7244 (2008)
Alessandro Di Maio et al.
EvoDevo, 6, 17-17 (2015-07-15)
Wnt signaling is one of the earliest and most highly conserved regulatory pathways for the establishment of the body axes during regeneration and early development. In regeneration, body axes determination occurs independently of tissue rearrangement and early developmental cues. Modulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service