Skip to Content
Merck
All Photos(1)

Documents

L6525

Sigma-Aldrich

Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H83N15O13
CAS Number:
Molecular Weight:
1090.28
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

product name

Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide, ≥97% (HPLC)

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CCCC[C@H](NC(=O)[C@H](C)NC(=O)[C@H](CCC(O)=O)NC(=O)[C@H](Cc1ccc(cc1)[N+]([O-])=O)NC(=O)[C@H](CCCC)NC(=O)[C@@H](NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](C)NC(=O)[C@@H](N)CCCCN)C(C)C)C(N)=O

InChI

1S/C49H83N15O13/c1-7-9-15-33(40(52)67)58-41(68)29(6)57-44(71)36(22-23-38(65)66)60-47(74)37(26-30-18-20-31(21-19-30)64(76)77)62-45(72)34(16-10-8-2)61-48(75)39(27(3)4)63-46(73)35(17-13-25-55-49(53)54)59-42(69)28(5)56-43(70)32(51)14-11-12-24-50/h18-21,27-29,32-37,39H,7-17,22-26,50-51H2,1-6H3,(H2,52,67)(H,56,70)(H,57,71)(H,58,68)(H,59,69)(H,60,74)(H,61,75)(H,62,72)(H,63,73)(H,65,66)(H4,53,54,55)/t28-,29-,32-,33-,34-,35-,36-,37-,39-/m0/s1

InChI key

MJGGTDHEIUOPEB-WBMHOGBOSA-N

Amino Acid Sequence

Lys-Ala-Arg-Val-Nle-pNO2-Phe-Glu-Ala-Nle-NH2

Application

Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide has been used for measuring HIV (human immunodeficiency virus)-1 protease activity.

Biochem/physiol Actions

Lys-Ala-Arg-Val-Nle-p-nitro-Phe-Glu-Ala-Nle amide is a synthetic substrate for HIV (human immunodeficiency virus)-1 protease and is commonly used for measuring HIV-1 protease activity.

Substrates

Substrate for HIV protease.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L H Phylip et al.
Biochemical and biophysical research communications, 171(1), 439-444 (1990-08-31)
Kinetic constants (Km,Kcat) are derived for the hydrolysis of a number of chromogenic peptide substrates by the aspartic proteinase from HIV-2. The effect of systematic replacement of the P2 residue on substrate hydrolysis by HIV-1 and HIV-2 proteinases is examined.
C Dash et al.
The Journal of biological chemistry, 276(4), 2487-2493 (2000-10-24)
The active site cleft of the HIV-1 protease (PR) is bound by two identical conformationally mobile loops known as flaps, which are important for substrate binding and catalysis. The present article reports, for the first time, an HIV-1 PR inhibitor
Anita Fehér et al.
The Journal of general virology, 87(Pt 5), 1321-1330 (2006-04-11)
The protease (PR) of Murine leukemia virus (MLV) was expressed in Escherichia coli, purified to homogeneity and characterized by using various assay methods, including HPLC-based, photometric and fluorometric activity measurements. The specificity of the bacterially expressed PR was similar to
Yunfeng Tie et al.
The FEBS journal, 272(20), 5265-5277 (2005-10-13)
HIV-1 protease (PR) and two drug-resistant variants--PR with the V82A mutation (PR(V82A)) and PR with the I84V mutation (PR(I84V))--were studied using reduced peptide analogs of five natural cleavage sites (CA-p2, p2-NC, p6pol-PR, p1-p6 and NC-p1) to understand the structural and
A D Richards et al.
The Journal of biological chemistry, 265(14), 7733-7736 (1990-05-15)
By replacement of the P1' residue in a capsid/nucleocapsid cleavage site mimic with 4-NO2-phenylalanine (Nph), an excellent chromogenic substrate, Lys-Ala-Arg-Val-Leu*Nph-Glu-Ala-Met, for HIV-1 proteinase (kappa cat = 20 s-1, Km = 22 microM) has been prepared. Substitution of the Leu residue

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service