Skip to Content
Merck
All Photos(1)

Key Documents

L3271

Sigma-Aldrich

Lys-Tyr-Lys acetate salt

≥97% (HPLC)

Synonym(s):

Lysyltyrosyllysine, acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H35N5O5 · xC2H4O2
Molecular Weight:
437.53 (free base basis)
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Assay

≥97% (HPLC)

form

powder

color

white to off-white

storage temp.

−20°C

SMILES string

CC(O)=O.NCCCC[C@H](N)C(=O)N[C@@H](Cc1ccc(O)cc1)C(=O)N[C@@H](CCCCN)C(O)=O

InChI

1S/C21H35N5O5.C2H4O2/c22-11-3-1-5-16(24)19(28)26-18(13-14-7-9-15(27)10-8-14)20(29)25-17(21(30)31)6-2-4-12-23;1-2(3)4/h7-10,16-18,27H,1-6,11-13,22-24H2,(H,25,29)(H,26,28)(H,30,31);1H3,(H,3,4)/t16-,17-,18-;/m0./s1

InChI key

OXXOYNFTOCTMGG-UVJOBNTFSA-N

Amino Acid Sequence

Lys-Tyr-Lys

Application

Lysyltyrosyllysine (Lys-Tyr-Lys, KYK) is used together with uracil DNA glycosylase to quantify uracil. Lysyltyrosyllysine is also used as a model peptide to study free radical oxidation and electron capture mechanisms.

Biochem/physiol Actions

Tripeptide can induce nicks at apurinic/apyrimidinic sites in circular DNA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

V Favaudon
International journal of radiation biology, 55(3), 353-364 (1989-03-01)
OH.-induced covalent peptide-nucleotide adducts have been isolated by reverse-phase chromatography from the enzymic hydrolyzates of gamma-ray irradiated solutions containing double-stranded poly(deoxyadenylic-deoxythymidylic acid) and one of the tripeptides, lysyl-tryptophyl-lysine or lysyl-tyrosyl-lysine. Numerous compounds were formed, resulting presumably from different modes of
J Pierre et al.
The Journal of biological chemistry, 256(20), 10217-10220 (1981-10-25)
Tripeptides containing aromatic residues between basic ones, such as Lys-Trp-Lys and Lys-Tyr-Lys can nick supercoiled or relaxed DNAs containing apurinic/apyrimidinic sites (AP-sites). The nicking, which is ionic strength-dependent, occurs at AP-sites, since native PM2 DNA is not hydrolyzed. The nicking
The effect of neighboring amino acid residues and solution environment on the oxidative stability of tyrosine in small peptides.
Zhang J, Kalonia DS.
AAPS PharmSci, 8, E102-E102 (2007)
James S Prell et al.
Journal of the American Chemical Society, 130(38), 12680-12689 (2008-09-03)
The effects of water on electron capture dissociation products, molecular survival, and recombination energy are investigated for diprotonated Lys-Tyr-Lys solvated by between zero and 25 water molecules. For peptide ions with between 12 and 25 water molecules attached, electron capture
R Barthwal et al.
Journal of molecular recognition : JMR, 4(2-3), 45-52 (1991-03-01)
The complete sequential assignment and conformation of d-GpCpGpC in D2O has been determined from 1D NMR spectra at 285-320 K and room temperature 2D-COSY and NOESY spectra. The tetradeoxynucleotide exists primarily as a right handed double helix at 285 K

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service