L1900
Luria broth base, Miller′s modified
Tryptone, yeast extract and sodium chloride
Synonym(s):
Lennox broth base, modified, Luria-Bertani broth base, modified, Lysogeny broth base, modified
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106200
NACRES:
NA.72
Recommended Products
form
powder
technique(s)
microbiological culture: suitable
application(s)
agriculture
microbiology
General description
Luria broth (LB) is a nutritionally rich bacterial culture medium that is commonly used for maintaining and cultivating laboratory recombinant strains of E. coli. It has potential applications in molecular microbiology methods such as the preparation of plasmid DNA and recombinant proteins. Luria broth base, Miller′s modified is used to culture members of the family Enterobacteriaceae and for coliphage plaque assays.
Application
Luria broth base, Miller′s modified has been used as a nutrient culture medium for E-coli cells.
Luria broth base (LB), Miller (L1900) is a bacteria growth mixture of sodium chloride (0.5g/L); tryptone (10g/L) and yeast extract (5g/L) typically combined with agar (15g/L).
Luria broth base (LB), Miller (L1900) is a bacteria growth mixture of sodium chloride (0.5g/L); tryptone (10g/L) and yeast extract (5g/L) typically combined with agar (15g/L).
Components
Contains: Tryptone, yeast extract and sodium chloride.
Reconstitution
Recommended use at 15.5 g per liter.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Facile monitoring and evaluation of bacteria in a fermentation process using perfusion chromatography and polarimetry detection
Berthod A, et al.
Journal of Separation Science, 26.0, 20-28 (2003)
Plasmid DNA topology assayed by two-dimensional agarose gel electrophoresis
Schvartzman JB, et al.
Journal of Separation Science, 121-132 (2013)
Plasmid DNA topology assayed by two-dimensional agarose gel electrophoresis
Schvartzman JB, et al.
Journal of Separation Science, 121-132 (2013)
Facile monitoring and evaluation of bacteria in a fermentation process using perfusion chromatography and polarimetry detection
Berthod A, et al.
Journal of Separation Science, 26(1-2), 20-28 (2003)
Facile monitoring and evaluation of bacteria in a fermentation process using perfusion chromatography and polarimetry detection
Berthod A, et al.
Journal of Separation Science, 26.0, 20-28 (2003)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service