The bead concentration is not determined on a lot to lot basis. However, historical data indicate a concentration of approximately 5.71 billion particles per milliliter.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
form
aqueous suspension
Quality Segment
composition
Solids, 2.5%
technique(s)
cell based assay: suitable
mean particle size
2.0 μm
fluorescence
λex ~360 nm; λem ~420 nm
application(s)
cell analysis
Application
Biochem/physiol Actions
1 of 1
This Item | |||
|---|---|---|---|
| description aqueous suspension, 2.0 μm mean particle size | description aqueous suspension, 0.05 μm mean particle size | description aqueous suspension, 1.0 μm mean particle size | description aqueous suspension, 2.0 μm mean particle size |
| technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) - |
| fluorescence λex ~360 nm; λem ~420 nm | fluorescence λex ~360 nm; λem ~420 nm | fluorescence λex ~470 nm; λem ~505 nm | fluorescence λex ~520 nm; λem ~540 nm |
| application(s) cell analysis | application(s) cell analysis | application(s) cell analysis | application(s) - |
| form aqueous suspension | form aqueous suspension | form aqueous suspension | form aqueous suspension |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| composition Solids, 2.5% | composition Solids, 2.4% | composition Solids, 2.5% | composition Solids, 2.4-2.6% |
Still not finding the right product?
Explore all of our products under Latex beads, amine-modified polystyrene, fluorescent blue
Storage Class
10 - Combustible liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the concentration of beads in 1 mL?
1 answer-
Helpful?
-



