Skip to Content
Merck

K3253

Kinetin solution

1 mg/mL, BioReagent, suitable for plant cell culture

Synonym(s):

6-Furfurylaminopurine, N6-Furfuryladenine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
NACRES:
NA.72
PubChem Substance ID:
UNSPSC Code:
10171502
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile-filtered

Quality Segment

product line

BioReagent

form

solution

concentration

1 mg/mL

technique(s)

cell culture | plant: suitable

application(s)

agriculture

shipped in

dry ice

storage temp.

−20°C

SMILES string

C(Nc1ncnc2[nH]cnc12)c3ccco3

InChI

1S/C10H9N5O/c1-2-7(16-3-1)4-11-9-8-10(13-5-12-8)15-6-14-9/h1-3,5-6H,4H2,(H2,11,12,13,14,15)

InChI key

QANMHLXAZMSUEX-UHFFFAOYSA-N

General description

Kinetin is a synthetic cytokinin, which functions as a plant growth regulator.

Application

Kinetin is an adenine-type cytokinin phytohormone that is used in plant culture media such as Murashige & Skoog media in conjunction with auxins such as indole-3-acetic acid (IAA), Indole-3-butyric acid (IBA), 1-napthaleneacetic acid (NAA) or others to induce the fomation of callus and to regenerate plant tissues from callus.

Biochem/physiol Actions

Kinetin is a type of cytokinin that is a plant hormone that promotes cell division. It is a growth factor that exerts several anti-aging and antioxidant effects on cells and organs. It is found to protect, effectively, the rat spleen from aging, apoptosis, and atrophy. It can help in inhibiting apoptosis, while promoting the proliferation of splenic lymphocyte, and effectively enhancing the immune power of the aging rats and slow down the aging process. It is also useful for enhancing phytoextraction.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library