Skip to Content
Merck
All Photos(3)

Key Documents

K2753

Sigma-Aldrich

(±)-Ketamine hydrochloride

≥99% (HPLC), solid, NMDA glutamate receptor antagonist

Synonym(s):

(±)-2-(2-Chlorophenyl)-2-(methylamino)cyclohexanone hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16ClNO · HCl
CAS Number:
Molecular Weight:
274.19
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Product Name

(±)-Ketamine hydrochloride, solid

form

solid

drug control

USDEA Schedule III; stupéfiant (France); regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain)

storage temp.

2-8°C

SMILES string

Cl.CNC1(CCCCC1=O)c2ccccc2Cl

InChI

1S/C13H16ClNO.ClH/c1-15-13(9-5-4-8-12(13)16)10-6-2-3-7-11(10)14;/h2-3,6-7,15H,4-5,8-9H2,1H3;1H

InChI key

VCMGMSHEPQENPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Selective NMDA glutamate receptor antagonist; veterinary anesthetic.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R V Pustovit et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 27(5), 610-617 (2015-01-24)
Despite constipation being a common problem, the treatments that are available have side effects and are only partly effective. Recent studies show that centrally penetrant ghrelin receptor agonists cause defecation in humans and other species. Here, we describe some features
Scott A Schobel et al.
Neuron, 78(1), 81-93 (2013-04-16)
The hippocampus in schizophrenia is characterized by both hypermetabolism and reduced size. It remains unknown whether these abnormalities are mechanistically linked. Here we addressed this question by using MRI tools that can map hippocampal metabolism and structure in patients and
Aude Castel et al.
BMC veterinary research, 9, 59-59 (2013-03-30)
Central post-stroke pain is a neuropathic pain condition caused by a vascular lesion, of either ischemic or hemorrhagic origin, in the central nervous system and more precisely involving the spinothalamocortical pathway responsible for the transmission of painful sensations. Few animal
Songen Zhang et al.
Journal of leukocyte biology, 97(6), 1003-1010 (2015-01-15)
Streptococcus pyogenes of the M1 serotype can cause STSS, which is associated with significant morbidity and mortality. The purpose of the present study was to examine the role of NFAT signaling in M1 protein-induced lung injury. NFAT-luc mice were treated
Yongzhi Wang et al.
Shock (Augusta, Ga.), 42(4), 343-349 (2014-07-01)
Systemic inflammatory response syndrome and severe infections are associated with major derangements in the coagulation system. The purpose of this study was to examine the dynamic alterations in thrombin generation in abdominal sepsis. Abdominal sepsis was induced by cecal ligation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service