Skip to Content
Merck
All Photos(1)

Documents

J4644

Sigma-Aldrich

Anti-phospho-JNK1/2 (pThr183/pTyr185) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-SAPK (pTpY183/185), Anti-phospho-Stress Activated Protein Kinase (pTpY183/185), Anti-phospho-c-Jun N-terminal Kinase 1/2 (pTpY183/185)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

rat, human

technique(s)

immunocytochemistry: 1:250
immunofluorescence: 1:250
immunohistochemistry: Assay Dependent
western blot: 1:200-1:2000 (using human 293 cells treated with UV radiation)

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... MAPK8(5599)

General description

c-Jun N-terminal kinase (JNK, SAPK) is an extensively studied memebr of the MAPK family of kinases and is critical in mediating the apoptotic signaling. There are three isoforms of JNK, JNK 1, 2 and 3. JNK 1 and 2 are ubiquitously expressed; JNK 3 is restricted to cardiac and neuronal tissues.

Immunogen

synthetic peptide derived from a region of JNK1 & 2 that contains threonine183 and tyrosine185.

Application

Anti-phospho-JNK1/2 (pThr183/pTyr185 ) antibody concentration is formulated to be used at the recommended dilutions by application.

Biochem/physiol Actions

JNK (c-Jun N-terminal kinase) is phosphorylated by MAPK (mitogen activated protein kinase) kinases 4 and 7 at Thr183 and Tyr185, leading to its activation. It is however not clear as to which MAPKK phosphorylates which amino acid residue. The activation of JNK is highly regulated and is in response to the stress signals that the cell faces. JNK phosphorylates c-Jun, the most important member of the AP-1 transcription factor family that is involved in survival, apoptosis, proliferation and tumorigenesis. JNK also activates other transcription factors such as c-myc and p53. JNK is involved in the phosphorylation of proteins such as Bad, Bcl2 and Bim that result in the execution of apoptosis by release of cytochrome c from mitochondria.
Anti-phospho-JNK1/2 (SAPK) (pThr183/pTyr185 ) specifically recognizes the endogenous, active forms of JNK1 and 2 phosphorylated at threonine 183 and tyrosine 185.

Physical form

Solution in Dulbecco′s phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, with 50% glycerol, 1% bovine serum albumin (BSA) (IgG and preservatives free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaurav Verma et al.
Journal of cellular physiology, 227(5), 1791-1795 (2011-07-07)
Apoptosis or programmed cell death is an extremely coordinated phenomenon that involves the participation of a complex interacting crosstalk between the endoplasmic reticulum and mitochondria. This involves a series of signaling molecules like stress kinases, caspases, Bcl-2 family of proteins
Anning Lin et al.
Aging cell, 1(2), 112-116 (2003-07-29)
Age-associated changes in apoptotic rates have been observed in a number of different tissues. While the implications of these findings remain unclear, a better understanding of how apoptosis is regulated may further our understanding of the aging process. The role
F Urano et al.
Science (New York, N.Y.), 287(5453), 664-666 (2000-01-29)
Malfolded proteins in the endoplasmic reticulum (ER) induce cellular stress and activate c-Jun amino-terminal kinases (JNKs or SAPKs). Mammalian homologs of yeast IRE1, which activate chaperone genes in response to ER stress, also activated JNK, and IRE1alpha-/- fibroblasts were impaired
Ksenya Cohen-Katsenelson et al.
The Biochemical journal, 439(3), 381-390 (2011-07-14)
JNK (c-Jun N-terminal kinase) is part of a MAPK (mitogen-activated protein kinase) signalling cascade. Scaffold proteins simultaneously associate with various components of the MAPK signalling pathway and play a crucial role in signal transmission and MAPK regulation. WDR62 (WD repeat
Jing Liu et al.
Cell research, 15(1), 36-42 (2005-02-03)
JNK is a key regulator of many cellular events, including programmed cell death (apoptosis). In the absence of NF-kB activation, prolonged JNK activation contributes to TNF-a induced apoptosis. JNK is also essential for UV induced apoptosis. However, recent studies reveal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service