Skip to Content
Merck
All Photos(1)

Key Documents

I9910

Sigma-Aldrich

Ibutilide hemifumarate salt

≥98% (HPLC)

Synonym(s):

(±)-N-[4-[4-(Ethylheptylamino)-1-hydroxybutyl]phenyl]-methanesulfonamide hemifumarate salt, Corvert

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H36N2O3S · 0.5C4H4O4
CAS Number:
Molecular Weight:
442.61
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

off-white to tan

solubility

H2O: >20 mg/mL

storage temp.

room temp

SMILES string

OC(=O)\C=C\C(O)=O.CCCCCCCN(CC)CCCC(O)c1ccc(NS(C)(=O)=O)cc1.CCCCCCCN(CC)CCCC(O)c2ccc(NS(C)(=O)=O)cc2

InChI

1S/2C20H36N2O3S.C4H4O4/c2*1-4-6-7-8-9-16-22(5-2)17-10-11-20(23)18-12-14-19(15-13-18)21-26(3,24)25;5-3(6)1-2-4(7)8/h2*12-15,20-21,23H,4-11,16-17H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

PCIOHQNIRPWFMV-WXXKFALUSA-N

Gene Information

human ... KCNH2(3757)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Ibutilide hemifumarate salt is considered a new generation "pure" Class III antiarrhythmic agent.
Ibutilide hemifumarate salt is considered a new generation "pure" Class III antiarrhythmic agent. Ibutilide binds to and alters the activity of hERG potassium channels, delayed inward rectifier potassium (IKr) channels and L-type (dihydropyridine sensitive) calcium channels.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jianling Sun et al.
Cell biochemistry and biophysics, 59(3), 153-158 (2010-10-26)
Ibutilide is a newer class-III antiarrhythmic agent approved for clinical use. We sought to investigate its electrophysiological effects in canines and also the underlying mechanism of conversion of atrial flutter (AFL). For this purpose, 15 male mongrel dogs were anesthetized
Jian-ling Sun et al.
Zhonghua xin xue guan bing za zhi, 37(10), 920-924 (2010-02-09)
To observe the electrophysiological effects of ibutilide on canine and to explore the potential mechanisms of ibutilide on terminating atrial flutter. Eighteen mongrel dogs were anesthetized and intubated. The heart was exposed through thoracotomy for electrodes implantation. The electrophysiologic variables
Efstathios D Pagourelias et al.
Cardiology, 122(2), 89-92 (2012-06-29)
We present a case of Brugada syndrome in a young patient whose typical ECG pattern was 'masked' after ibutilide was administered for atrial flutter cardioversion. Ibutilide, a class III antiarrhythmic agent used for the treatment of atrial fibrillation and flutter
Atrial fibrillation: AVRO--were the results a surprise?
Soraya M Samii et al.
Nature reviews. Cardiology, 8(4), 188-190 (2011-03-03)
Predicting the degree of drug-induced QT prolongation and the risk for torsades de pointes.
Chunhua Ding
Heart rhythm, 8(10), 1535-1536 (2011-06-28)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service