I8757
4-Iodo-L-phenylalanine
≥98.0% (TLC)
Synonym(s):
(S)-2-Amino-3-(4-iodophenyl)propanoic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C9H10INO2
CAS Number:
Molecular Weight:
291.09
Beilstein:
4411317
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
Recommended Products
Product Name
4-Iodo-L-phenylalanine,
Assay
≥98.0% (TLC)
Quality Level
form
powder
color
white to off-white
storage temp.
−20°C
SMILES string
N[C@@H](Cc1ccc(I)cc1)C(O)=O
InChI
1S/C9H10INO2/c10-7-3-1-6(2-4-7)5-8(11)9(12)13/h1-4,8H,5,11H2,(H,12,13)/t8-/m0/s1
InChI key
PZNQZSRPDOEBMS-QMMMGPOBSA-N
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
4-Iodo-L-phenylalanine may be used in protein engineering as a model unnatural α amino acid to alter primary amino acid composition via the opal (UGA) codon.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
R Karthikraj et al.
Rapid communications in mass spectrometry : RCM, 26(11), 1385-1391 (2012-05-05)
In chiral differentiation by mass spectrometry, use of a single reference that differentiates various classes of compounds including drugs is ideal, but so far there are no such reports in the literature. We have successfully used iodo-substituted amino acids for
Veerle Kersemans et al.
European journal of nuclear medicine and molecular imaging, 33(8), 919-927 (2006-03-31)
In vitro in the R1M cell model and in vivo in the R1M tumour-bearing athymic model, both [(123)I]-2-iodo-L: -phenylalanine and [(123)I]-2-iodo-D: -phenylalanine have shown promising results as tumour diagnostic agents for SPECT. In order to compare these two amino acid
B Chevrier et al.
European journal of biochemistry, 237(2), 393-398 (1996-04-15)
The structure of the complex of Aeromonas proteolytica aminopeptidase, a two-zinc exopeptidase, with the inhibitor p-iodo-D-phenylalanine hydroxamate has been determined by X-ray crystallography. Refinement of the structure, which includes 220 water molecules, using data at 0.80-0.23-nm resolution resulted in a
Koichiro Kodama et al.
Journal of biochemistry, 148(2), 179-187 (2010-05-25)
A variety of unique codons have been employed to expand the genetic code. The use of the opal (UGA) codon is promising, but insufficient information is available about the UGA suppression approach, which facilitates the incorporation of non-natural amino acids
Koichiro Kodama et al.
Chembiochem : a European journal of chemical biology, 7(10), 1577-1581 (2006-09-14)
An Escherichia coli suppressor tRNA(Phe) (tRNA(Phe) (CUA)) was misacylated with 4-iodo-L-phenylalanine by using the A294G phenylalanyl-tRNA synthetase mutant (G294-PheRS) from E. coli at a high magnesium-ion concentration. The preacylated tRNA was added to an E. coli cell-free system and a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service