Skip to Content
Merck

I8657

Interferon α from rat

≥95%, recombinant, expressed in bacteria, lyophilized powder, suitable for cell culture

Synonym(s):

IFN-α

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

rat

recombinant

expressed in bacteria

assay

≥95%

form

lyophilized powder

mol wt

~19 kDa

packaging

pkg of 1 vial

concentration

300,000-800,000,000 units/mL

technique(s)

cell culture | mammalian: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

General description

Interferon α is a cytokine, that belongs to type I interferon (IFNs) family. It possesses 14 genes, including 2 pseudo-genes or allelic forms that encodes for 13 proteins. It is mainly induced in response to viral infection. It is a non-leukemogenic peptide.

Biochem/physiol Actions

Cytokine with anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities.
Cytokine with anti-viral, anti-proliferative, immunoregulatory, and proinflammatory activities. Type I Interferons are a closely related family of 165-172 amino acid proteins that are produced by leukocytes (α subtypes), fibroblasts (β subtypes), lymphocytes (ω subtypes) and ruminant embryos (τ subtypes). Interferon induces a variety of effects on target cells including antiviral, antiproliferative and immunomodulatory activities.
Interferon α play a key role in the inhibition of in vitro proliferation of hematopoietic progenitors. It mediates the regulation of gene expression that is responsible for development and functionality of innate and adaptive immunity. It has been used as myelosuppressive agent for myeloproliferative neoplasms (MPNs).

Physical form

Lyophilized from a solution containing citric acid buffered saline (CBS pH 5.0) and 125 mM trehalose

Analysis Note

The biological activity is determined in the cytopathic effect inhibition assay using rat epithelial-like (RATEC) cells with vesicular stomatitis virus (VSV) in the presence of 2mM glutathione. The cytopathic effect of 50% is produced with ~1 unit/mL interferon. Since there is no international rat interferon α standard, units are determined with respect to the NIH international standard reference for mouse interferon α/β.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library