Skip to Content
Merck

I7763

Mini ice tub

capacity 1 L

Synonym(s):

188481102, M18848-1102, benchtop ice bucket, cooling receptical, receptical, small ice bucket

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41103000
Manufacturer/tradename:
Bel-Art M18848-1102
Packaging:
pack of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


packaging

pack of 1 ea

manufacturer/tradename

Bel-Art M18848-1102

L

3 in.

capacity

1 L

top L × W

6 in. × 6 in.

General description

One-piece expanded PVC, offering chemical resistance (acids, acetone, chloroform, etc.) and thermal resistance for wet ice, dry ice, and liquid nitrogen.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library