Skip to Content

I5403

Insulin-Like Growth Factor Binding Protein-2 human

recombinant, expressed in NSO cells, lyophilized powder, suitable for cell culture, ≥95% (SDS-PAGE)

Synonym(s):

IGFBP-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51111800
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in NSO cells

assay

≥95% (SDS-PAGE)

form

lyophilized powder

potency

0.04 μg/mL

mol wt

~36 kDa by SDS-PAGE, calculated mol wt ~31 kDa

packaging

pkg of 25 μg

storage condition

avoid repeated freeze/thaw cycles

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... IGFBP2(3485)

General description

The gene IGFBP-2 (insulin-like growth factor binding protein-2) is mapped to human chromosome 2q35.

Biochem/physiol Actions

IGFBP-2 (insulin-like growth factor binding protein-2) regulates the function of IGF ligands. IGFBP2 is associated with skeletal muscle cell proliferation and differentiation. It also plays an important role in the differentiation of corneal fibroblast.
During development, insulin-like growth factor binding protein-2 is expressed in a number of tissues with the highest expression level found in the central nervous system. IGFBP-2 exhibits a 2-10 fold higher affinity for IGF II than for IGF I. Mouse and human IGFBP-2 share approximately 82% homology.
Exhibits a 2-10 fold higher affinity for IGF II than for IGF I.

Physical form

Lyophilized from a 0.2 μm filtered solution of 30% (v/v) acetonitrile with 0.1% (v/v) TFA

Analysis Note

The biological activity is measured by its ability to inhibit recombinant human lGF-II on MCF-7 cells.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library