Skip to Content
Merck
All Photos(4)

Documents

I3636

Sigma-Aldrich

Acridine Mutagen ICR 191

Synonym(s):

6-Chloro-9-[3-(2-chloroethylamino)propylamino]-2-methoxyacridine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21Cl2N3O · 2HCl
CAS Number:
Molecular Weight:
451.22
Beilstein:
8176205
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

solubility

DMSO: soluble

suitability

suitable for Ames test in Salmonella and E. coli (Frameshift mutagenic standard)

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].COc1ccc2nc3cc(Cl)ccc3c(NCCCNCCCl)c2c1

InChI

1S/C19H21Cl2N3O.2ClH/c1-25-14-4-6-17-16(12-14)19(23-9-2-8-22-10-7-20)15-5-3-13(21)11-18(15)24-17;;/h3-6,11-12,22H,2,7-10H2,1H3,(H,23,24);2*1H

InChI key

LMEMIKWTNPWYMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 2 Dermal - Acute Tox. 2 Oral - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K F Kiser et al.
Oncogene, 25(50), 6595-6603 (2006-05-17)
Retroviral integration mutagenesis and treatment with the frameshift mutagen ICR191 were used to transform v-H-ras expressing PB-3c cells to interleukin-3 (IL-3) independence. Six clones displayed viral integrations into the 3' region of the IL-3 gene thus acting post-transcriptionally by disrupting
Katerina Vlasakova et al.
Mutation research, 578(1-2), 225-237 (2005-09-07)
A novel forward mutation assay has been developed in Salmonella typhimurium based on resistance to 5-fluorouracil (FU). The mutational target in the FU assay was determined to be the uracil phosphoribosyl transferase (upp) gene. To validate the upp gene as
S R Sahasrabudhe et al.
Genetics, 129(4), 981-989 (1991-12-01)
As the most nucleophilic site in DNA, the guanine N7 atom is a major site of adduction by a large number of alkylating mutagens and carcinogens. Aflatoxin B1, a powerful mutagen, is believed to act through its reaction with this
Kathryn Annett et al.
Experimental gerontology, 40(12), 976-981 (2005-10-12)
T cells undergo rapid clonal expansion upon antigenic stimulation to produce an effective immune response. Any defect in the DNA mismatch repair (MMR) system may have a detrimental effect on T cell proliferation. This study employed an in vitro model
Richard Hampson et al.
Mutation research, 525(1-2), 1-9 (2003-03-26)
Mutagenesis screening, in which heritable traits are isolated following damage to the genome, is a powerful approach for investigating gene function. Among vertebrate model organisms, the zebrafish (Danio rerio) is ideally suited to mutagenesis screens. The success of large-scale screens

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service