Skip to Content
Merck
All Photos(4)

Key Documents

I2886

Sigma-Aldrich

3-Indoleacetic acid

suitable for plant cell culture, crystalline

Synonym(s):

indole acetic acid, Heteroauxin, IAA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO2
CAS Number:
Molecular Weight:
175.18
Beilstein:
143358
EC Number:
MDL number:
UNSPSC Code:
10171502
PubChem Substance ID:
NACRES:
NA.72
Pricing and availability is not currently available.

Assay

≥98%

Quality Level

form

crystalline

technique(s)

cell culture | plant: suitable

color

white to pink

mp

165-169 °C (lit.)

application(s)

agriculture

storage temp.

−20°C

SMILES string

OC(=O)Cc1c[nH]c2ccccc12

InChI

1S/C10H9NO2/c12-10(13)5-7-6-11-9-4-2-1-3-8(7)9/h1-4,6,11H,5H2,(H,12,13)

InChI key

SEOVTRFCIGRIMH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Indoleacetic acid has been used:
  • as standard to quantify hormones[1]
  • to modify MS (Murashige and Skoog) media for the initiation of callus[2]
  • as an external standard in the analyses of free endogenous growth regulators[3]

Indole-3-acetic acid (IAA) is a natural phytohormone (plant auxin) used as a supplement in media such as Murashige and Skoog media and Gamborg′s B5 media. IAA is a plant signaling molecule involved in plant organogenesis and growth control.

Preparation Note

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Free endogenous growth regulators in Pistachio (Pistacia vera L.)
Okay Y, et al.
African Journal of Agricultural Research, 6(5), 1161-1169 (2011)
Callus induction and organogenesis in soybean [Glycine max (L.) Merr.] cv. Pyramid from mature cotyledons and embryos
Joyner EY, et al.
The Open Plant Science Journal, 4(1) (2010)
Auxin, gibberellin, cytokinin and abscisic acid production in some bacteria
Karadeniz A, et al.
World Journal of Microbiology & Biotechnology, 22(10), 1061-1064 (2006)
Xu Liu et al.
PLoS biology, 17(4), e3000219-e3000219 (2019-04-12)
Macroautophagy (hereafter autophagy) is a well-conserved cellular process through which cytoplasmic components are delivered to the vacuole/lysosome for degradation and recycling. Studies have revealed the molecular mechanism of transcriptional regulation of autophagy-related (ATG) genes upon nutrient deprivation. However, little is
Xin Wen et al.
Autophagy, 16(7), 1172-1185 (2019-08-30)
Macroautophagy/autophagy, a highly conserved dynamic process, is one of the major degradative pathways in cells. So far, over 40 autophagy-related (ATG) genes have been identified in Saccharomyces cerevisiae, most of which have homologs in more complex eukaryotes. Autophagy plays a

Protocols

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Reference guide and preparation guide for antibiotic and antimycotic use in plant tissue culture.

Questions

  1. I want to use this IAA to induce human OsTIR1-Auxin-inducible-degron cell line to degrade protein of interest. If this product is appliable in this experiment.

    1 answer
    1. This product is tested for use in plant tissue culture. Suitability for other applications would have to be determined experimentally by the end user. This compound is soluble in ethanol at 50 mg/mL and may be diluted further with medium or buffer. Please see the links below to review publications that may be helpful:
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC7659001/
      https://www.ncbi.nlm.nih.gov/pmc/articles/PMC7544234/

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service