Skip to Content
Merck

I2002

Isocitric Dehydrogenase (NADP) from porcine heart

Type IV, buffered aqueous glycerol solution, 3-20 units/mg protein

Synonym(s):

threo-Ds-Isocitrate: NADP+ oxidoreductase, ICDH, IDH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
200-289-5
MDL number:
EC Number:
Specific activity:
3-20 units/mg protein
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type IV

form

buffered aqueous glycerol solution

quality

≤ 0.2% of main activity is obtained with NAD as cofactor.

specific activity

3-20 units/mg protein

composition

Protein biuret

shipped in

wet ice

storage temp.

2-8°C

Application

Isocitric dehydrogenase (IDH) has been used:
  • for in vitro activity assay to study the effects of thorium on citric acid cycle enzymes in the mitochondrial respiration pathway
  • as a standard to measure the impact of temperature on nicotinamide diphosphate (NADP)-IDH activity
  • in isocitrate assay

Biochem/physiol Actions

Isocitrate dehydrogenase (IDH) is a homodimeric enzyme and participates in several processes such as adapting to hypoxia, histone demethylation, and DNA modification. This enzyme has three isoforms like IDH1, IDH2, and IDH3.

Physical form

Solution in 50% glycerol in EDTA buffer salts, pH 6.0

Other Notes

One unit will convert 1.0 μmole of isocitrate to α-ketoglutarate per min at pH 7.4 at 37 °C.


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Icksoo Lee et al.
Biochimica et biophysica acta, 1802(2), 275-283 (2009-10-20)
Noonan syndrome (NS) is an autosomal dominant disorder, and a main feature is congenital heart malformation. About 50% of cases are caused by gain-of-function mutations in the tyrosine phosphatase SHP2/PTPN11, a downstream regulator of ERK/MAPK. Recently it was reported that
Avtar Singh et al.
The Journal of biological chemistry, 278(35), 33208-33216 (2003-06-07)
Transhydrogenase couples the reduction of NADP+ by NADH to inward proton translocation across mitochondrial and bacterial membranes. The coupling reactions occur within the protein by long distance conformational changes. In intact transhydrogenase and in complexes formed from the isolated, nucleotide-binding
Sergiy M Nadtochiy et al.
The Journal of biological chemistry, 291(38), 20188-20197 (2016-08-12)
2-Hydroxyglutarate (2-HG) is an important epigenetic regulator, with potential roles in cancer and stem cell biology. The d-(R)-enantiomer (d-2-HG) is an oncometabolite generated from α-ketoglutarate (α-KG) by mutant isocitrate dehydrogenase, whereas l-(S)-2-HG is generated by lactate dehydrogenase and malate dehydrogenase



Global Trade Item Number

SKUGTIN
I2002-25MG04061832730448
I2002-250MG04061832730431
I2002-5G-A04061832490205
I2002-25G-A04061833848241
I2002-100MG04061833848234
I2002-500MG04061832730455