Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
I1146
R3 IGF-1 human
recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Synonym(s):
Human IGF-1, IGF-1 Protein, Recombinant IGF-1
About This Item
Recommended Products
biological source
human
Quality Level
recombinant
expressed in E. coli
Assay
≥85% (HPLC)
form
lyophilized powder
potency
≤10 ng/mL ED50
quality
endotoxin tested
mol wt
7,676 Da
packaging
pkg of 0.1 mg
technique(s)
cell culture | mammalian: suitable
UniProt accession no.
storage temp.
2-8°C
Gene Information
human ... IGF1(3479)
Application
Recombinantly produced in E. coli using a patented expression system, the peptide is harvested from cells in inclusion bodies, which are dissolved and desalted. The active molecule is properly folded under oxidizing conditions and then purified by several liquid chromatography steps.
Biochem/physiol Actions
Other Notes
Preparation Note
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Insulin-like Growth Factors (IGF)
-
What is the Department of Transportation shipping information for this product?
1 answer-
Helpful?
-
-
How do the sequences of Products I1146 and I1271 compare to that of native Insulin-like Growth Factor 1 (IGF-1)?
1 answer-
The entire IGF-1 gene codes for 194 amino acids, but the mature form of the protein is 70 amino acids, consisting of amino acids 49-118.Product I1146 is R3 IGF-1, which is also 70 amino acids in length. The Glutamic acid (E) at position 3 has simply been replaced by an Arginine (R). So the sequence of Product I1146 is:GP RTLCGAELVD ALQFVCGDRG FYFNKPTGYG SSSRRAPQTG IVDECCFRSC DLRRLEMYCA PLKPAKSAProduct I1271 is the same as I1146, but with an additional thirteen amino acids "in front of" the normal N-terminus of the R3 IGF-1:MFPAM PLSSLFVNGP RTLCGAELVD ALQFVCGDRG FYFNKPTGYG SSSRRAPQTG IVDECCFRSC DLRRLEMYCA PLKPAKSANote that the first 13 amino acids in the recombinant I1271 (MFPAMPLSSLFVN)are quite different from what would have been the preceding 13 amino acids in the immature IGF-1.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service