Skip to Content

HT90232

Gram′s Iodine Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.47
UNSPSC Code:
41116121
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label

IVD

for in vitro diagnostic use

application(s)

hematology
histology
microbiology

storage temp.

room temp

Application

Used as a kit component in Gram Stain Procedure (Sigma Procedure HT90).

Other Notes

Iodine, 0.33%, and potassium iodide, 0.66%.


Still not finding the right product?

Explore all of our products under Gram′s Iodine Solution


flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Repr. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library