Skip to Content
Merck

HT40332

Papanicolaou Stain EA 50

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116121
NACRES:
NA.47
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solution

Quality Segment

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

technique(s)

microbe id | staining: suitable

application(s)

hematology
histology

storage temp.

room temp

General description

For routine diagnostic cytology to aid in the identification and classification of exfoliative cells.

Application

Papanicolaou stain imparts a characteristic range of coloration to exfoliative cells of vaginal, cervical, prostatic and other body secretions, allowing critical examination of nuclei and cytoplasmic components.

Other Notes

Eosin Y (certified), 0.23%,Bismarck brown (certified), 0.05%, fast green FCF (certified), 0.08%, phosphotungstic acid, 0.2%, in denatured alcohol


Still not finding the right product?

Explore all of our products under Papanicolaou Stain EA 50


signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 2

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

55.4 °F - closed cup

flash_point_c

13 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library