Merck
HS29025F
Microtube Rack
orange rack
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific HS29025F
Feature:
to hold 80 tube(s)
Material:
orange rack, polypropylene
Packaging:
pack of 5 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
orange rack, polypropylene
feature
to hold 80 tube(s)
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS29025F
L × W × H
226 mm × 67 mm × 28 mm
General description
The 80-Well Microtube Rack is a popular fraction-collector-plate style rack.
Features and Benefits
  • Easily fill and transfer 80 tubes in compact arrangement (5x16 array)
  • Frosted labeling area on bottom of rack
  • Embossed Index Numbers for quick sample ID and retrieval
  • Made of polypropylene
  • Autoclavable
Still not finding the right product?
Explore all of our products under Microtube Rack
Choose from one of the most recent versions:
Certificates of Analysis (COA)