Merck
HS2344A
Reversible PCR Tube Rack
to hold, 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes, assorted colors polypropylene
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.12
UNSPSC Code:
41122800
Manufacturer/tradename:
Heathrow Scientific HS2344A
Feature:
to hold 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes
Material:
assorted colors polypropylene
Packaging:
pack of 5 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
assorted colors polypropylene
feature
to hold 168 x 0.2 mL/ 40 x 0.5 mL/12 x 1.5-2.0 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
Heathrow Scientific HS2344A
L × W × H
206 mm × 131 mm × 54 mm
General description
The PCR Racks are reversible and allow you to use 0.2,0.5, and 1.5/2.0mL tubes. The removable hinged lid fits both sides of rack.
Choose from one of the most recent versions:
Certificates of Analysis (COA)