Merck
HS23325A
Arctic Squares® Cryostorage Box
Holds 25 x 2 mL tubes, red polycarbonate, pk of 8
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.22
UNSPSC Code:
41123304
Manufacturer/tradename:
Heathrow Scientific HS23325A
Feature:
Holds 25 x 2 mL tubes
Material:
red polycarbonate
Packaging:
pk of 8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
red polycarbonate
feature
Holds 25 x 2 mL tubes
packaging
pk of 8
manufacturer/tradename
Heathrow Scientific HS23325A
technique(s)
cryopreservation: suitable
size
76x76x53 mm
General description
Safely stores vials from -196 to 121C in mechanical freezers as well as liquid nitorgen. Boxes are designed with unique features to help orient, identify and access vials with ease.
  • forward sloped base and high contrast indexing on the transparent lid ensure quick visual orientation
  • vend and drainage holes
  • stackable
  • autoclavable
Features and Benefits
  • ideal for small sampling
  • friction fit, slide on lid
Legal Information
Arctic Squares is a registered trademark of Heathrow Scientific LLC
Still not finding the right product?
Explore all of our products under Arctic Squares® Cryostorage Box
Choose from one of the most recent versions:
Certificates of Analysis (COA)