Skip to Content

HS1423BB

Weigh Boat

Medium, black polystyrene (square), Anti-static, pk of 500 ea

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41121812
Material:
black polystyrene (square)
Packaging:
pk of 500 ea
Manufacturer/tradename:
Heathrow Scientific HS1423BB
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black polystyrene (square)

size

medium

feature

Anti-static

packaging

pk of 500 ea

manufacturer/tradename

Heathrow Scientific HS1423BB

L × W × H

(85 mm) × 84 mm × 24 mm

color

black

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.

Features and Benefits

  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer



Still not finding the right product?

Explore all of our products under Weigh Boat




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library