Skip to Content
Merck

Skip To

HS1420BB

Weigh Boat

Medium, square white polystyrene, Anti-Static, pack of 500 ea

Synonym(s):

weigh boat, weigh dish, weighing dish, weighing pan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.71
UNSPSC Code:
41121812
Material:
square white polystyrene
Packaging:
pack of 500 ea
Manufacturer/tradename:
Heathrow Scientific HS1420BB
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

square white polystyrene

size

medium

feature

Anti-Static

packaging

pack of 500 ea

manufacturer/tradename

Heathrow Scientific HS1420BB

L × W × H

85 mm × 85 mm × 24 mm

color

white

General description

Flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.

Features and Benefits

  • biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
  • temperature range -10 to 70C
  • disposable
  • shallow with a wide, flat bottom to resist tipping
  • rounded corners simplify transfer


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HS1420CHS1420BHS120223
description

Medium, square white polystyrene, Anti-Static, pack of 500 ea

description

Large, square white polystyrene, pack of 500 ea

description

Medium, square white polystyrene, pack of 500 ea

description

Medium, square blue polystyrene, Anti-static, pk of 500 ea

manufacturer/tradename

Heathrow Scientific HS1420BB

manufacturer/tradename

Heathrow Scientific HS1420C

manufacturer/tradename

Heathrow Scientific HS1420B

manufacturer/tradename

Heathrow Scientific HS120223

material

square white polystyrene

material

square white polystyrene

material

square white polystyrene

material

square blue polystyrene

packaging

pack of 500 ea

packaging

pack of 500 ea

packaging

pack of 500 ea

packaging

pk of 500 ea

color

white

color

white

color

white

color

blue

feature

Anti-Static

feature

-

feature

-

feature

Anti-static

L × W × H

85 mm × 85 mm × 24 mm

L × W × H

140 mm × 140 mm × 22 mm

L × W × H

85 mm × 85 mm × 24 mm

L × W × H

(85 mm) × 84 mm × 24 mm


Still not finding the right product?

Explore all of our products under Weigh Boat




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library