Skip to Content
Merck

HS120252

Wide mouth safety labelled wash bottle

capacity 500 mL, label, Isopropanol, self venting, PE-LD

Synonym(s):

Heathrow Scientific HS120252, cleaning bottle, dispensing bottle, drip bottle, drop bottle, fluid dispenser, laboratory wash bottle, solvent wash bottle, squeezable bottle, washbottle

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.43
UNSPSC Code:
41103209
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

PE-LD , low-density polyethylene bottle, polypropylene cap, wide-mouth bottle, yellow cap

joint

joint threaded

feature

label Isopropanol, vented, with cap

packaging

pack of 6 ea

manufacturer/tradename

Heathrow Scientific HS120252

capacity

500 mL

General description

The information provided on the front of each bottle is specific to the product contained and includes the following:
  • Chemical Abstract Service (CAS) No.
  • National Fire Protection Association (NFPA) codes and symbols
  • Protective Equipment Symbol (PES)
  • Target Organ Information and Hazard Codes.
You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling’ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the floSelf Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE

Application

Used for easy identification of solutions and solvents for use on the bench.

Features and Benefits

  • Wide bottle mouth makes filling easier
  • Non-metallic vent that prevents the risk of solvent leakage
  • Closure and stem are molded in one piece to make them leak-proof
  • Bottles are made from LDPE, with a PP closure with the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection
  • Six named chemicals, a self labeling and assorted pack of bottles to choose from
  • 500 mL bottle size
  • Supplied in packs of 6


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library