Skip to Content
Merck
All Photos(6)

Key Documents

HPA051870

Sigma-Aldrich

Anti-TMEM119 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Tmem119 Antibody, Tmem119 Antibody - Anti-TMEM119 antibody produced in rabbit, Anti-transmembrane protein 119

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

GDGARMVEGRGAEEEEKGSQEGDQEVQGHGVPVETPEAQEEPCSGVLEGAVVAGEGQGELEGSLLLAQEAQGPVGPPESPCACSSVHPS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Transmembrane protein 119 (TMEM119) is encoded by the gene mapped to human chromosome 12q23.3. The encoded protein belongs to the transmembrane protein family. TMEM119 has an O-glycosylated N-terminal region.

Immunogen

transmembrane protein 119 recombinant protein epitope signature tag (PrEST)

Application

Anti-TMEM119 antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Transmembrane protein 119 (TMEM119) acts as an osteoinductive factor and facilitates proliferation, migration and invasion of osteosarcoma cells.(28} It is a critical molecule acting downstream of the parathyroid hormone (PTH) and similar to mothers against decapentaplegic-3 (Smad3) signaling pathways in osteoblasts. TMEM119 is considered to be a potential microglial marker that differentiates resident microglia from blood-derived macrophages in the human brain. Mutation in the gene is associated with the development of osteosarcoma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST85802

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tobias Zrzavy et al.
Brain : a journal of neurology, 140(7), 1900-1913 (2017-05-26)
Microglia and macrophages accumulate at the sites of active demyelination and neurodegeneration in the multiple sclerosis brain and are thought to play a central role in the disease process. We used recently described markers to characterize the origin and functional
Parathyroid hormone-responsive Smad3-related factor, Tmem119, promotes osteoblast differentiation and interacts with the bone morphogenetic protein-Runx2 pathway.
Hisa I, et al.
The Journal of Biological Chemistry, 286(11), 9787-9796 (2011)
Alexander J Ehrenberg et al.
Journal of neuroscience methods, 339, 108708-108708 (2020-04-04)
Neurodegenerative diseases feature stereotypical deposits of protein aggregates that selectively accumulate in vulnerable cells. The ability to simultaneously localize multiple targets in situ is critical to facilitate discovery and validation of pathogenic molecular pathways. Immunostaining methods enable in situ detection
Upregulation and biological function of transmembrane protein 119 in osteosarcoma.
Jiang Z H, et al.
Experimental & Molecular Medicine, 49(5), e329-e329 (2017)
Involvement of the osteoinductive factors, Tmem119 and BMP-2, and the ER stress response PERK?eIF2??ATF4 pathway in the commitment of myoblastic into osteoblastic cells.
Tanaka K I, et al.
Calcified Tissue International, 94(4), 454-464 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service