Skip to Content
Merck
All Photos(5)

Key Documents

HPA044659

Sigma-Aldrich

Anti-CD47 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-IAP, Anti-MER6, Anti-OA3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

AQLLFNKTKSVEFTFCNDTVVIPCFVTNMEAQNTTEVYVKWKFKGRDIYTFDGALNKSTVPTDFSSAKIEVSQLLKGDASLKMDKSDAVSHTGNYTCEVTELTREGETIIELKYRVVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD47(961)

General description

Cluster of differentiation 47 (CD47) is also termed as integrin-associated protein (IAP). It is a glycoprotein that belongs to the immunoglobulin superfamily. CD47 is a highly glycosylated, ~50 kDa membrane protein. It has a single IgV-like domain at its N-terminus, a highly hydrophobic stretch with five membrane-spanning segments and an alternatively spliced cytoplasmic C-terminus. CD47 is located on human chromosome 3q13.

Immunogen

CD47 molecule

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry. The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-CD47 antibody has been used in immunohistochemical analysis.

Biochem/physiol Actions

Cluster of differentiation 47 (CD47) plays a crucial role in self-recognition. It acts as an inhibitor of phagocytosis with the help of ligation of signal regulatory protein that are expressed on phagocytes. Overexpression of CD47 results in increased risk of tumor growth and metastasis. It participates in β3 integrin-mediated signaling on leukocytes and also controls cell migration, axon extension, cytokine production and T cell activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86204

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD47 expression for in situ and invasive cutaneous epithelial lesions.
Randa Akel et al.
Journal of the American Academy of Dermatology, 75(2), 434-436 (2016-07-23)
Lesions referred to dermatology in the Department of Veterans Affairs (VA) health system: A retrospective chart review.
Katherine R Grey et al.
Journal of the American Academy of Dermatology, 75(2), 430-434 (2016-07-23)
Tumor-selective Blockade of CD47 Signaling with CD47 Antibody for Enhanced Antitumor Activity in Malignant Meningioma.
Liu, et al.
Current Neuropharmacology (2023)
Is CD47 an innate immune checkpoint for tumor evasion?.
Liu X, et al.
Journal of Hematology & Oncology, 10(1), 12-12 (2017)
CD47 expression for in situ and invasive cutaneous epithelial lesions.
Akel R, et al.
Journal of the American Academy of Dermatology, 75(2), 434-436 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service