Skip to Content
Merck
All Photos(4)

Key Documents

HPA039980

Sigma-Aldrich

Anti-NRP2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Neuropilin 2, Anti-Vegf165r2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

LSLTFHTDMAVAKDGFSARYYLVHQEPLENFQCNVPLGMESGRIANEQISASSTYSDGRWTPQQSRLHGDDNGWTPNLDSNKEYL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NRP2(8828)

General description

The gene NRP2 (neuropilin 2) is mapped to human chromosome 2q33. The gene encodes a single-spanning transmembrane glycoprotein. It belongs to the NRP class of proteins. The protein has two CUB (complement C1r/C1s, Uegf, Bmp1) domains, two F5/8 domains, a MAM (meprin/A5 protein/receptor protein tyrosine phosphatase μ) domain and a transmembrane region.

Immunogen

neuropilin 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NRPs (neuropilins) mainly interact with vascular endothelial growth factor (VEGF) receptors, class 3 semaphorins and other receptors. Absence of NRP2 in mice shows mild phenotypic effects in lymphatic system development and axon guidance. NRP2 is also expressed in various cancers and is associated with the proliferation, survival and migration of cancer cells. NRP2 interaction with SEMA3F (semaphorin 3F) reduces PI-3K (phosphoinositide 3 kinase) activity, mTORC2 (mammalian target of rapamycin complex 2)-associated signaling, RhoA (Ras homolog gene family, member A) activity and suppresses the making of cytoskeletal stress fibers.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78075

Physical form

Solution in phosphate buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

USCAP 2022 Abstracts: Genitourinary Pathology (including renal tumors) (522-659).
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 35(Suppl 2), 657-806 (2022-03-19)
USCAP 2022 Abstracts: Genitourinary Pathology (including renal tumors) (522-659).
Laboratory investigation; a journal of technical methods and pathology, 102(Suppl 1), 583-732 (2022-03-19)
Concurrent Expression of VEGF-C and Neuropilin-2 Is Correlated with Poor Prognosis in Glioblastoma.
Zhao H, et al.
The Tohoku Journal of Experimental Medicine, 238, 85-91 (2016)
Sequence Requirements for Neuropilin-2 Recognition by ST8SiaIV and Polysialylation of Its O-Glycans.
Bhide GP, et al.
The Journal of Biological Chemistry, 291, 9444-9457 (2016)
Triggers for autism: genetic and environmental factors.
Matsuzaki H, et al.
Journal of Central Nervous System Disease, 4, 27-36 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service