Skip to Content
Merck
All Photos(1)

Key Documents

HPA031131

Sigma-Aldrich

Anti-KCNV2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Kv8.2, Anti-potassium channel, subfamily V, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500-1:1000

immunogen sequence

WNTTENEGSQHRRSICSLGARSGSQASIHGWTEGNYNYYIEEDEDGEEEDQWKDDLAEEDQQAGEVTTAKPEGPSDPPALLSTLNVNVGGHSYQLDYCELA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KCNV2(169522)

General description

The KCNV2 (potassium channel, subfamily V, member 2) gene is mapped to human chromosome 9p24.2. KCNV2 is highly expressed in retinal photoreceptors. It encodes a modulatory subunit of the Kv8.2, voltage-gated potassium channel.

Immunogen

potassium channel, subfamily V, member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

KCNV2 (potassium channel, subfamily V, member 2) significantly influences the initiation and shaping of action potentials. KCNV2 mutations might lead to febrile, afebrile partial seizures, and epileptic encephalopathy. Alterations in KCVN2 gene might cause an abnormality in photoreceptor membrane potentials by inhibiting proper assembly of heteromeric voltage-gated potassium channels following a pathologically sustained outward potassium current in the dark. Mutation in the gene causes cone dystrophy with supernormal rod responses.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73048

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of cis- and trans-acting genetic variants explaining up to half the variation in circulating vascular endothelial growth factor levels.
Debette S
Circulation Research, 109(5), 554-563 (2011)
Molecular characteristics of four Japanese cases with KCNV2 retinopathy: report of novel disease-causing variants.
Fujinami K
Molecular Vision, 19, 1580-1590 (2013)
Pharmacogenetic and case-control study on potassium channel related gene variants and genetic generalized epilepsy.
Qu J
Medicine, 96(26) (2017)
Novel compound heterozygous mutations resulting in cone dystrophy with supernormal rod response.
Lenis TL
JAMA Ophthalmology, 131(11), 1482-1485 (2013)
Jacqueline Gayet-Primo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(14), 3414-3427 (2018-02-28)
Silent voltage-gated potassium channel subunits (KVS) interact selectively with members of the KV2 channel family to modify their functional properties. The localization and functional roles of these silent subunits remain poorly understood. Mutations in the KVS subunit, KV8.2 (KCNV2), lead

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service