Skip to Content
Merck
All Photos(4)

Documents

HPA027241

Sigma-Aldrich

Anti-ECM1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Extracellular matrix protein 1, Anti-Secretory component p85

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

EDTLDKYCDREYAVKTHHHLCCRHPPSPTRDECFARRAPYPNYDRDILTIDIGRVTPNLMGHLCGNQRVLTKHKHIPGLIHNMTARCCDLPFPEQACCAEEEKLTFINDLCGPRRNIWRDPALCCYLSPGDEQVNCFNINYLRNVALVS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ECM1(1893)

General description

Extracellular matrix protein 1 (ECM1) is a glycoprotein. It is expressed in skin and other tissues. The gene encoding this protein is localized on human chromosome 1q21.2.

Immunogen

Extracellular matrix protein 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-ECM1 antibody produced in rabbit has been used in immunofluorescence staining.

Biochem/physiol Actions

Extracellular matrix protein 1 (ECM1) has a role in cell proliferation, mineralization, and angiogenesis. The protein has been shown to have a role in cancer invasion and progression. It is associated with hepatocellular carcinoma. Mutations in the ECM1 gene has been linked to lipoid proteinosis. The protein also promotes the Warburg effect, which is an oncogenic switch that allows cancerous cells to take in more glucose than non-cancerous cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70702

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Extracellular matrix protein 1 gene (ECM1) mutations in nine Iranian families with lipoid proteinosis
Farzad Izadi
The Indian Journal of Medical Research (2016)
Valen Zhuoyou Yu et al.
Cancer letters, 461, 56-64 (2019-07-19)
Esophageal squamous cell carcinoma (ESCC) is a deadly disease with dismal 5-year survival. Extracellular matrix protein 1 (ECM1) was identified as one of the most downregulated genes by transcriptomic analysis of normal esophageal/ESCC paired tissue samples. ECM1 plays oncogenic roles
ECM1 promotes migration and invasion of hepatocellular carcinoma by inducing epithelial-mesenchymal transition
Hao Cheng
World Journal of Surgical Oncology (2016)
ECM1 promotes the Warburg effect through EGF-mediated activation of PKM2.
Lee KM
Cellular Signalling (2015)
Sophie Sarah Steinhaeuser et al.
Laboratory investigation; a journal of technical methods and pathology, 100(7), 928-944 (2020-03-24)
The tumor microenvironment is increasingly recognized as key player in cancer progression. Investigating heterotypic interactions between cancer cells and their microenvironment is important for understanding how specific cell types support cancer. Forming the vasculature, endothelial cells (ECs) are a prominent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service