Skip to Content
Merck
All Photos(1)

Key Documents

HPA020130

Sigma-Aldrich

Anti-SLC12A2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-BSC, Anti-BSC2, Anti-NKCC1, Anti-PPP1R141

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

TLVLGFKKDWLQADMRDVDMYINLFHDAFDIQYGVVVIRLKEGLDISHLQGQEELLSSQEKSPGTKDVVVSVEYSKKSDLDTSKPLSE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC12A2(6558)

General description

Solute carrier family 12 member 2 (SLC12A2) is a member of the cation–chloride cotransporters (CCCs) family. The SLC12A2 gene is mapped to human chromosome 5q23.3. It is widely expressed in neurons and also in other organs especially, in the secretory epithelial cells. SLC12A2 exists as eight isoforms. It comprises 12 transmembrane (TM) helices and assembles into a dimer with TM11-TM12 helices lining the dimer interface.

Immunogen

Solute carrier family 12 member 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-SLC12A2 antibody produced in rabbit has been used in immunofluorescence (1:1000).

Biochem/physiol Actions

Solute carrier family 12 member 2 (SLC12A2) also called Na-K-Cl cotransporter sodium-potassium-chloride cotransporter 1 (NKCC1) is a neuronal chloride importer and a neuronal chloride (Cl−) accumulator. It functions to regulate intraneuronal chloride levels along with KCC2. Mutations in the SLC12A2 are implicated in impaired secretory epithelia, encephalopathy, and neurodevelopmental disorders(NDD). It may serve as a potential target to treat neuropathic disorders, pain, and seizures.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74676

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alisdair McNeill et al.
Brain : a journal of neurology, 143(8), 2380-2387 (2020-07-14)
The SLC12 gene family consists of SLC12A1-SLC12A9, encoding electroneutral cation-coupled chloride co-transporters. SCL12A2 has been shown to play a role in corticogenesis and therefore represents a strong candidate neurodevelopmental disorder gene. Through trio exome sequencing we identified de novo mutations
Tommy Stödberg et al.
Neurology. Genetics, 6(4), e478-e478 (2020-08-06)
To describe the phenotype in 2 sisters with a rare constellation of neurologic symptoms and secretory impairments and to identify the etiology by the use of whole-genome sequencing (WGS). After an extensive workup failed to reveal the cause of disease
Angela-Maria Merli et al.
Journal of Crohn's & colitis (2020-09-14)
Ulcerative colitis (UC) patients have a greater risk of developing colorectal cancer through inflammation-dysplasia-carcinoma sequence of transformation. The histopathological diagnosis of dysplasia is therefore of critical clinical relevance, but dysplasia may be difficult to distinguish from inflammatory changes. A proteomic
Giulia Gorrieri et al.
Scientific reports, 6, 36016-36016 (2016-10-28)
Goblet cell hyperplasia, a feature of asthma and other respiratory diseases, is driven by the Th-2 cytokines IL-4 and IL-13. In human bronchial epithelial cells, we find that IL-4 induces the expression of many genes coding for ion channels and
Xiaoyong Yang et al.
Nature communications, 11(1), 1016-1016 (2020-02-23)
The secondary active cation-chloride cotransporters (CCCs) utilize the existing Na+ and/or K+ gradients to move Cl- into or out of cells. NKCC1 is an intensively studied member of the CCC family and plays fundamental roles in regulating trans-epithelial ion movement

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service