Skip to Content
Merck
All Photos(7)

Key Documents

HPA019467

Sigma-Aldrich

Anti-TAGLN antibody produced in rabbit

enhanced validation

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

TAGLN Antibody - Anti-TAGLN antibody produced in rabbit, Tagln Antibody, Anti-22 kDa actin-binding protein, Anti-SM22-alpha, Anti-Smooth muscle protein 22-alpha, Anti-Transgelin, Anti-WS3-10

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

EKKYDEELEERLVEWIIVQCGPDVGRPDRGRLGFQVWLKNGVILSKLVNSLYPDGSKPVKVPENPPSMVFKQMEQV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TAGLN(6876)

General description

The gene TAGLN (transgelin) is mapped to human chromosome 11q23.2. It belongs to the calponin family. The protein is strongly expressed in the smooth muscle tissues.

Immunogen

Transgelin recombinant protein epitope signature tag (PrEST)

Application

Anti-TAGLN antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

TAGLN (transgelin) is an actin stress fiber binding protein. It is mainly involved in cell growth, differentiation, invasion, muscle fiber contractility and matrix remodeling. TAGLN is up-regulated in osteosarcoma cell lines, colorectal adenocarcinoma, prostate , hepatocellular , gastric, pancreatic and colon cancer. However, it is down-regulated in urinary bladder, renal cell carcinoma and colorectal cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74706

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Deepthi Rao et al.
Human pathology, 46(6), 876-883 (2015-04-07)
Triple negative (TN) (estrogen receptor [ER], progesterone receptor [PR] and HER2-) are highly aggressive, rapidly growing, hormone-unresponsive tumors diagnosed at later stage that affect younger women with shorter overall survival. Most TN tumors are of the basal type. For the
Mingdong Zhao et al.
International journal of molecular medicine, 34(6), 1565-1572 (2014-10-17)
Alterations in the expression of microRNAs (miRNAs or miRS) have been implicated in the pathogenesis of the majority of human malignancies, and the dysregulation of microRNA-144 (miR-144) has been associated with several diseases. However, the potential involvement of miR-144 in
Ben Davidson et al.
Gynecologic oncology, 128(2), 349-355 (2012-11-28)
Endometrial stromal sarcoma (ESS) and leiomyosarcoma (LMS) are the two most common uterine sarcomas, but both are rare tumors. The aim of the present study was to compare the global gene expression patterns of ESS and LMS. Gene expression profiles
Naïma Kaci-Ouchfoun et al.
Asian journal of andrology, 12(3), 422-430 (2010-04-20)
The seminal vesicles of adult sand rat contain a major secretory protein band (MW 21 kDa) designated as Psammomys obesus seminal vesicles protein of 21 kDa (POSVP(21)). This protein is abundant in secretions, regulated by androgens and also present in
Gun-Hoo Park et al.
Oncology reports, 32(4), 1347-1354 (2014-08-12)
Neurofibromatosis type 1 (NF1) caused by NF1 gene mutation is a commonly inherited autosomal dominant disorder. Malignant peripheral nerve sheath tumors (MPNSTs), a type of aggressive sarcoma, are a major cause of mortality in NF1 patients. The malignant transformation of benign plexiform

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service