Skip to Content
Merck
All Photos(2)

Documents

HPA019000

Sigma-Aldrich

Anti-PHLDA1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Apoptosis-associated nuclear protein, Anti-PQ-rich protein, Anti-Pleckstrin homology-like domain family A member 1, Anti-Proline- and glutamine-rich protein, Anti-Proline- and histidine-rich protein, Anti-T-cell death-associated gene 51 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200
western blot: 0.4 μg/mL

immunogen sequence

RMLESSGCKALKEGVLEKRSDGLLQLWKKKCCILTEEGLLLIPPKQLQHQQQQQQQQQQQQQQPGQGPAEPSQPSGPAVASLEPPVKLKELHFSNMKTVDCVE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHLDA1(22822)

General description

The gene PHLDA1 (pleckstrin homology-like domain family A member 1) is mapped to human chromosome 12q15. It belongs to pleckstrin homology-related domain family. PHLDA1 is widely expressed. The protein is present in the cytoplasm and nucleolus. PHLDA1 is also referred as TDAG51 (T-cell death-associated gene 51).

Immunogen

Pleckstrin homology-like domain family A member 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

In activated T lymphocytes, PHLDA1 (pleckstrin homology-like domain family A member 1) works with T-cell receptor to inhibit protein biosynthesis. It is also responsible for differentiation of trichoepithelioma from basal cell carcinomas (BCCs). PHLDA1 is down-regulated in primary, and metastatic melanomas and breast cancer. On the other hand, it is involved with migration and proliferation in colon cancer cells. PHLDA1 is also involved in development of atherosclerosis in presence of hyperhomocysteinemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74199

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maxime Battistella et al.
The American Journal of dermatopathology, 36(8), 643-650 (2013-05-31)
Granular-cell or clear-cell basal cell carcinomas (BCCs), and clear-cell trichoblastomas have rarely been reported in the literature. PHLDA1 is a follicular stem cell marker, the expression of which has been reported to differentiate trichoepithelioma from BCCs. We wondered whether (1)
Assignment of the human PHLDA1 gene to chromosome 12q15 by radiation hybrid mapping.
M D Kuske et al.
Cytogenetics and cell genetics, 89(1-2), 1-1 (2000-07-15)
T Hinz et al.
Cellular signalling, 13(5), 345-352 (2001-05-23)
The T cell death associated gene 51 (TDAG51) was shown to be required for T cell receptor (TCR)-dependent induction of Fas/Apo1/CD95 expression in a murine T cell hybridoma. Despite the absence of a nuclear localization sequence and a nucleic acid
Emmanuel O Johnson et al.
Journal of cell science, 124(Pt 16), 2711-2722 (2011-08-03)
Aurora A kinase is overexpressed in the majority of breast carcinomas. A chemical genetic approach was used to identify the malignant targets of Aurora A, which revealed pleckstrin-homology-like domain protein PHLDA1 as an Aurora A substrate. PHLDA1 downregulation is a
Rüdiger Neef et al.
Cancer research, 62(20), 5920-5929 (2002-10-18)
To identify molecules involved in the progression of human melanoma to metastatic disease, autologous primary and metastatic melanoma cells were compared by differential mRNA display. One cDNA, expressed in primary but not in autologous metastatic cells in three different patients

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service