Skip to Content
Merck
All Photos(7)

Documents

HPA018002

Sigma-Aldrich

Anti-SIGMAR1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Aging-associated gene 8 protein, Anti-OPRS1, Anti-SIG-1R, Anti-SR-BP, Anti-SR31747-binding protein, Anti-Sigma 1-type opioid receptor, Anti-Sigma1-receptor, Anti-Sigma1R, Anti-hSigmaR1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

FVFQREEIAQLARQYAGLDHELAFSRLIVELRRLHPGHVLPDEELQWVFVNAGG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... OPRS1(10280)

General description

Sigma non-opioid intracellular receptor 1 (SIGMAR1) is a 223-amino acid protein that is present in the endoplasmic reticulum and plasma membrane. The gene encoding it is localized on chromosome 9p13.1-p13.3.

Immunogen

Sigma 1-type opioid receptor recombinant protein epitope signature tag (PrEST)

Application

Anti-SIGMAR1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Sigma non-opioid intracellular receptor 1 (SIGMAR1) acts as a ligand-regulated molecular chaperone. It interacts with ion channels and G-protein coupled receptors. Studies have shown that SIGMAR1 inhibits the generation of reactive oxygen species (ROS) in the liver, lungs and retina. Mutations in the gene encoding SIGMAR1 have been associated with juvenile amyotrophic lateral sclerosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72457

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simon Couly et al.
Human molecular genetics, 29(4), 529-540 (2019-11-07)
Sigma-1 receptor (S1R) is an endoplasmic reticulum (ER) chaperone that not only regulates mitochondrial respiration but also controls cellular defense against ER and oxidative stress. This makes S1R a potential therapeutic target in amyotrophic lateral sclerosis (ALS). Especially, as a
Laura R Rodríguez et al.
Redox biology, 37, 101762-101762 (2020-11-01)
Friedreich ataxia (FRDA) is a neurodegenerative disorder characterized by neuromuscular and neurological manifestations. It is caused by mutations in the FXN gene, which results in loss of the mitochondrial protein frataxin. Endoplasmic Reticulum-mitochondria associated membranes (MAMs) are inter-organelle structures involved
Ilaria Genovese et al.
Cell death & disease, 11(10), 861-861 (2020-10-17)
Dysregulation of calcium signaling is emerging as a key feature in the pathogenesis of neurodegenerative diseases such as Alzheimer's disease (AD), Parkinson's disease (PD), and Huntington's disease (HD), and targeting this process may be therapeutically beneficial. Under this perspective, it
Xiaobo Li et al.
Neurology, 84(24), 2430-2437 (2015-06-17)
To identify the underlying genetic cause in a consanguineous Chinese family segregating distal hereditary motor neuropathy (dHMN) in an autosomal recessive pattern. We used whole-exome sequencing and homozygosity mapping to detect the genetic variant in 2 affected individuals of the
Amr Al-Saif et al.
Annals of neurology, 70(6), 913-919 (2011-08-16)
Amyotrophic lateral sclerosis (ALS) is a neurodegenerative disorder characterized by loss of motor neurons in the brain and spinal cord, leading to muscle weakness and eventually death from respiratory failure. ALS is familial in about 10% of cases, with SOD1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service