Skip to Content
Merck
All Photos(5)

Key Documents

HPA017742

Sigma-Aldrich

Anti-DDC antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-AADC, Anti-Aromatic-L-amino-acid decarboxylase, Anti-DDC, Anti-DOPA decarboxylase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human, mouse, rat

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

LETVMMDWLGKMLELPKAFLNEKAGEGGGVIQGSASEATLVALLAARTKVIHRLQAASPELTQAAIMEKLVAYSSDQAHSSVERAGLIGGVKLKAIPSDGNFAMRASA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DDC(1644)

General description

Aromatic-L-amino-acid decarboxylase (DDC) which is a pyridoxal 5′-phosphate dependent enzyme, is present in the cytosol and cell membrane. The enzymatically active form of the enzyme is detected in many tissues. The gene encoding DDC is localized on chromosome 7p11-p13.

Immunogen

Aromatic-L-amino-acid decarboxylase recombinant protein epitope signature tag (PrEST)

Application

Anti-DDC antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Aromatic-L-amino-acid decarboxylase (DDC) is an enzyme involved in the production of the neurotransmitters like dopamine and serotonin. It catalyzes the decarboxylation of 3, 4-dihydroxy-L-phenylalanine (L-Dopa) and L-5-hydroxytryptophan (5-HTP) to form dopamine and serotonin respectively. DDC also uses catechol- or indole-related L-amino acids. This enzyme is a coregulator of the androgen receptor (AR) in prostate cells and enhances its activity. Deficiency of DDC results in severe developmental delay and autonomic dysfunction. It has also been shown that the enzyme is involved in Parkinson′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72511

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Ewald et al.
Psychiatric genetics, 5(4), 161-169 (1995-01-01)
Dopa decarboxylase (DDC) is involved directly in the synthesis of dopamine and serotonin and indirectly in the synthesis of noradrenaline. The present study investigated two Danish families for linkage between manic depressive illness, a marker at the DDC locus which
Riccardo Montioli et al.
BioMed research international, 2013, 161456-161456 (2013-06-20)
Human Dopa decarboxylase (hDDC), a pyridoxal 5'-phosphate (PLP) enzyme, displays maxima at 420 and 335 nm and emits fluorescence at 384 and 504 nm upon excitation at 335 nm and at 504 nm when excited at 420 nm. Absorbance and
Panagiota-Aikaterini Geomela et al.
BMC cancer, 12, 484-484 (2012-10-23)
Head and neck squamous cell carcinoma (HNSCC) represents one of the most commonly diagnosed malignancies worldwide. The DDC gene encodes L-DOPA decarboxylase, an enzyme catalyzing the decarboxylation of L-DOPA to dopamine. We have recently shown that DDC mRNA is a
R Pons et al.
Neurology, 62(7), 1058-1065 (2004-04-14)
Deficiency of aromatic L-amino acid decarboxylase (AADC) is associated with severe developmental delay, oculogyric crises (OGC), and autonomic dysfunction. Treatment with dopamine agonists and MAO inhibitors is beneficial, yet long-term prognosis is unclear. To delineate the clinical and molecular spectrum
Riccardo Montioli et al.
Human molecular genetics, 23(20), 5429-5440 (2014-05-29)
Dopa decarboxylase (DDC), or aromatic amino acid decarboxylase (AADC), is a pyridoxal 5'-phosphate enzyme responsible for the production of the neurotransmitters dopamine and serotonin. Deficit of this enzyme causes AADC deficiency, an inherited neurometabolic disorder. To date, 18 missense homozygous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service