Skip to Content
Merck
All Photos(10)

Key Documents

HPA016903

Sigma-Aldrich

Anti-CD46 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD46 antigen, Anti-Membrane cofactor protein precursor, Anti-TLX, Anti-Trophoblast leukocyte common antigen

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
WB
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200-1:500
western blot: 0.04-0.4 μg/mL

immunogen sequence

NGKHTFSEVEVFEYLDAVTYSCDPAPGPDPFSLIGESTIYCGDNSVWSRAAPECKVVKCRFPVVENGKQISGFGKKFYYKATVMFECDKGFYLDGSDTIVCDSNSTWDPPVPKCLK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD46(4179)

General description

CD46 (CD46 molecule, complement regulatory protein) is a membrane cofactor protein, and behave as a complement regulatory receptor for C3b and other pathogens.

Immunogen

Membrane cofactor protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-CD46 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

CD46 (CD46 molecule, complement regulatory protein) is involved in the T-cell-mediated immunity and tolerance. In presence of IL-2, CD46 acts as a costimulatory factor and induces signals for the Tr1 (T-regulatory 1)-specific cytokine phenotype in human CD4+ T cells. These stimulated CD4+ cells multiply fast to restrict the activation of bystander T cells. It also plays a major role in human spermatozoon-oocyte interaction at the oocyte plasma membrane.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71847

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raine Toivonen et al.
Virology journal, 9, 296-296 (2012-11-30)
Clinical gene therapy trials for cardiovascular diseases have demonstrated the crucial role of efficient gene delivery and transfection technologies in achieving clinically relevant results. We hypothesized that the use of tropism-modified adenoviruses would improve transduction efficacy and to this end
A Astier et al.
Journal of immunology (Baltimore, Md. : 1950), 164(12), 6091-6095 (2000-06-08)
The widely expressed transmembrane molecule CD46 is the complement regulatory receptor for C3b as well as the receptor for several pathogens. Beside its binding functions, CD46 is also able to transduce signals. We showed that CD46 aggregation on human T
C T Taylor et al.
Human reproduction (Oxford, England), 9(5), 907-911 (1994-05-01)
CD46 (membrane cofactor protein) is a cell surface complement regulatory glycoprotein that facilitates enzymatic cleavage of complement component C3b; it is expressed by both human oocytes and acrosome-reacted spermatozoa. Murine anti-CD46 monoclonal antibody (mAb) has been reported to decrease significantly
Claudia Kemper et al.
Nature, 421(6921), 388-392 (2003-01-24)
The immune system must distinguish not only between self and non-self, but also between innocuous and pathological foreign antigens to prevent unnecessary or self-destructive immune responses. Unresponsiveness to harmless antigens is established through central and peripheral processes. Whereas clonal deletion

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service