Skip to Content
Merck
All Photos(5)

Key Documents

HPA015128

Sigma-Aldrich

Anti-B4GALNT1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GALGT, Anti-SPG26, Anti-beta1-4GalNAc-T

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

EVTGVTLTGEGQADLTLVSPGLDQLNRQLQLVTYSSRSYQTNTADTVRFSTEGHEAAFTIRIRHPPNPRLYPPGSLPQGAQYNISALVTIATKTFLRYDRLRALITSIRRFYPTVTVVIADDSDKPERVSGPYV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... B4GALNT1(2583)

Looking for similar products? Visit Product Comparison Guide

General description

B4GALNT1 (β-1,4-N-acetyl-galactosaminyl transferase 1) is a GalNAc transferase, and is also called GM2 synthase. This gene is located on human chromosome 12q. Two subunits, linked by disulphide bonds, form a B4GALNT1 homodimer. This protein is transmembrane in nature. Cathepsin D-like protease cleaves this enzyme, in the late Golgi complex, to produce a soluble form.

Immunogen

Beta-1,4 N-acetylgalactosaminyltransferase 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

B4GALNT1 (β-1,4-N-acetyl-galactosaminyl transferase 1) enzyme catalyses the second step in the biosynthesis of complex gangliosides, such as GM (mono)2, GA (asialo)2 and GD (di)2. Gangliosides are essential components of synaptic plasma membrane, and are essential for the development of central nervous system. Mutations in this gene are linked to the pathogenesis of hereditary spastic paraplegia. It is also expressed in small cell lung carcinoma (SCLC) and neuroblastoma cells. The presence of this protein in post-therapy bone marrow of neuroblastoma patients is related to poor prognosis and survival. Studies in mice show that inactivation of this gene leads to male sterility, as well as degeneration of axons, and suppressed myelination in peripheral and central nervous system.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71972

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Jaskiewicz et al.
The Journal of biological chemistry, 271(42), 26395-26403 (1996-10-18)
Many Golgi membrane-bound glycosyltransferases are released from cells in a soluble form. To characterize this release process, we stably transfected Chinese hamster ovary cells with three myc epitope-tagged forms of cloned beta1, 4-N-acetylgalactosaminyltransferase (GalNAcT); two of these forms resided in
Gaurav V Harlalka et al.
Brain : a journal of neurology, 136(Pt 12), 3618-3624 (2013-10-10)
Glycosphingolipids are ubiquitous constituents of eukaryotic plasma membranes, and their sialylated derivatives, gangliosides, are the major class of glycoconjugates expressed by neurons. Deficiencies in their catabolic pathways give rise to a large and well-studied group of inherited disorders, the lysosomal
Amir Boukhris et al.
American journal of human genetics, 93(1), 118-123 (2013-06-12)
Hereditary spastic paraplegias (HSPs) form a heterogeneous group of neurological disorders. A whole-genome linkage mapping effort was made with three HSP-affected families from Spain, Portugal, and Tunisia and it allowed us to reduce the SPG26 locus interval from 34 to
Lin-Chi Chen et al.
Lung cancer (Amsterdam, Netherlands), 67(2), 216-220 (2009-05-22)
GD2/GM2 synthase is a key enzyme in the synthesis of GD2 and GM2 gangliosides found on the surface of neuroblastoma and small cell lung carcinoma (SCLC) cells. In neuroblastoma, persistent levels of GD2/GM2 synthase RNA in bone marrow (BM) following
J Li et al.
The Journal of biological chemistry, 275(52), 41476-41486 (2000-10-06)
GM2 synthase is a homodimer in which the subunits are joined by lumenal domain disulfide bond(s). To define the disulfide bond pattern of this enzyme, we analyzed a soluble form by chemical fragmentation, enzymatic digestion, and mass spectrometry and a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service