Skip to Content
Merck
All Photos(4)

Documents

HPA014750

Sigma-Aldrich

Anti-LAMP1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD107a antigen, Anti-LAMP-1, Anti-Lysosome-associated membrane glycoprotein 1 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

SPVPKSPSVDKYNVSGTNGTCLLASMGLQLNLTYERKDNTTVTRLLNINPNKTSASGSCGAHLVTLELHSEGTTVLLFQFGMNASSSRFFLQGIQLNTILPDARDPAFKAANGSLRALQATVGNSYKCN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LAMP1(3916)

General description

LAMP1 (lysosomal-associated membrane protein 1) is a transmembrane lysosomal glycoprotein. The predicted molecular weight of this protein is 283kDa. It is composed of 385 amino acids, and has ~18 putative N-glycosylation sites. These sites are separated by a serine-proline rich region, into two domains. This region has similarity to immunoglobulin (Ig)A hinge domain. One N-glycosylated domain is present at the N-terminal, and is extended to a putative signal sequence, and the other domain is connected to the transmembrane region, and projects into the cytosol.

Immunogen

Lysosome-associated membrane glycoprotein 1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The exact function of LAMP1 (lysosomal-associated membrane protein 1) is not yet known. It might be involved in cell adhesion. This protein, in association with galectin, is involved in the differentiation of keratinocytes of epidermis. It is up-regulated in cancer cells, and is thought to facilitate tumor invasion, by regulating binding to endothelial cells and extracellular matrix (ECM). It is part of the particulate matrix, and is responsible for the induction of atrioventricular endothelium for its conversion into mesenchyme. This conversion occurs in heart in the atrioventricular canal and proximal outflow tract.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72362

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Viitala et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(11), 3743-3747 (1988-06-01)
Although several lysosomal membrane glycoproteins have been characterized by using specific antibodies, none of the studies so far elucidated the amino acid sequence of a lysosomal membrane glycoprotein. Here we describe cDNA clones encoding for one of the lysosome-associated membrane
Allan R Sinning et al.
The anatomical record. Part A, Discoveries in molecular, cellular, and evolutionary biology, 277(2), 307-311 (2004-03-31)
The heart extracellular matrix protein hLAMP-1 (lectin-associated matrix protein in the heart) is a component of the particulate matrix that activates the AV endothelium prior to its transformation into mesenchyme within the atrioventricular canal and proximal outflow tract of the
Victoria Sarafian et al.
Archives of dermatological research, 298(2), 73-81 (2006-05-20)
Lysosomes and their components are suspected to be involved in epidermal differentiation. In this study, lysosomal enzyme activities, expression of the lysosome-associated membrane protein 1 (Lamp-1) and expression of the epidermal galectins-1, -3 and -7 were investigated in human keratinocytes
Lukasz Kuźbicki et al.
Melanoma research, 16(3), 235-243 (2006-05-24)
Lysosome-associated membrane protein-1 is a protein with a significant content of beta1,6-branched N-glycans. It is thought that enhanced expression of lysosome-associated membrane protein-1 in tumour cells may promote invasion by influencing both adhesion to extracellular matrix and perhaps also binding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service