Skip to Content
Merck
All Photos(5)

Documents

HPA010592

Sigma-Aldrich

Anti-CD74 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CD74 antigen antibody produced in rabbit, Anti-HLA class II histocompatibility antigen γ-chain antibody produced in rabbit, Anti-HLA-DR antigens-associated invariant chain antibody produced in rabbit, Anti-Ia antigen-associated invariant chain antibody produced in rabbit, Anti-Ii antibody produced in rabbit, Anti-p33 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

PKPVSKMRMATPLLMQALPMGALPQGPMQNATKYGNMTEDHVMHLLQNADPLKVYPPLKGSFPENLRHLKNTMETIDWKVFESWMHHWLLFEMSRHSLEQKPTDAPPK

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD74(972)

General description

CD74 (cluster of differentiation 74) is the invariant chain of MHC (major histocompatibility) complex class II. It was the first receptor to be recognized for macrophage migration inhibitory factor (MIF). It is a membrane receptor.

Immunogen

HLA class II histocompatibility antigen γ-chain recombinant protein epitope signature tag (PrEST)

Application

Anti-CD74 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CD74 (cluster of differentiation 74) acts as the chaperone of MHC-II (major histocompatibility) and is responsible for the trafficking of MHC-II molecules in APCs (antigen presenting cells). It also interacts with non-MHC proteins and functions as a receptor for MIF (macrophage migration inhibitory factor). MIF is a pleiotropic cytokine having pro-tumorigenic and pro-inflammatory functionality. It is involved in the progression and metastasis of hematologic and solid cancers, and its up-regulation in thyroid cancer might be linked with advanced tumor stage. In patients with malignant pleural mesothelioma, the over-expression of this protein functions as an independent prognostic marker for overall survival.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71660

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Otterstrom et al.
British journal of cancer, 110(8), 2040-2046 (2014-03-07)
The pro-inflammatory cytokine migration inhibitory factor (MIF) and its receptor CD74 have been proposed as possible therapeutic targets in several cancers. We studied the expression of MIF and CD74 together with calretinin in specimens of malignant pleural mesothelioma (MPM), correlating
Marta Olah et al.
Nature communications, 11(1), 6129-6129 (2020-12-02)
The extent of microglial heterogeneity in humans remains a central yet poorly explored question in light of the development of therapies targeting this cell type. Here, we investigate the population structure of live microglia purified from human cerebral cortex samples
Cintia D'Amato-Brito et al.
Oncotarget, 7(10), 11512-11525 (2016-02-18)
Macrophage migration inhibitory factor (MIF) is a pro-inflammatory cytokine implicated in acute and chronic inflammatory diseases. MIF is overexpressed in various tumors. It displays a number of functions that provide a direct link between the process of inflammation and tumor
Dariusz Adamek et al.
Folia neuropathologica, 47(3), 240-246 (2009-10-09)
The expression of protein RCAS1 (receptor-binding cancer antigen expressed on SiSo cells), possibly involved in the mechanisms of evasion of immune surveillance by tumours, has been studied in brain astrocytomas grade III and IV and in metastatic carcinomas to the
Luisa Klemke et al.
Cell death & disease, 12(2), 155-155 (2021-02-06)
Macrophage migration inhibitory factor (MIF) is an upstream regulator of innate immunity, but its expression is increased in some cancers via stabilization with HSP90-associated chaperones. Here, we show that MIF stabilization is tumor-specific in an acute colitis-associated colorectal cancer (CRC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service