Skip to Content
Merck
All Photos(6)

Key Documents

HPA010547

Sigma-Aldrich

Anti-NFKBIZ antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-FLJ34463, Anti-INAP, Anti-MAIL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

QGRRALSYVLARKMNALHMLDIKEHNGQSAFQVAVAANQHLIVQDLVNIGAQVNTTDCWGRTPLHVCAEKGHSQVLQAIQKGAVGSNQFVDLEATNYDGLTPLHCAVIAHNAVVHELQRNQQPHSPEVQELLL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKBIZ(64332)

General description

NFKBIZ (NFKB inhibitor ξ), also known as IκBξ, is a member of the IκB inhibitor protein family. It is a nuclear protein, and is induced in macrophages and lung primary cells, following TLR/IL-1 (toll like receptor/ interleukin-1) signaling. Two mRNA of this protein is produced through alternate splicing, long and short, where the short form lacks the 99-amino acid sequence at the N-terminal, present in the long form.

Immunogen

NF-kappa-B inhibitor zeta recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NFKBIZ (NFKB inhibitor ζ) functions as an inhibitor of NFκB, and is controlled by it through negative feedback loop. It is also essential for the induction of certain inflammation-related gene such as IL6 (interleukin 6). Studies show that various variants of this gene are linked with susceptibility to invasive pneumococcal disease (IPD) in humans. This gene is regulated by IL1 and interferon (IFN)-γ, which in turn induces β-glucan-activated human dendritic cell programming. It also influences both canonical and non-canonical NF?B signaling in activated B-cell-like (ABC) subtype of diffuse large B-cell lymphoma (DLBCL).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71497

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Cardone et al.
PloS one, 9(12), e114516-e114516 (2014-12-05)
Recognition of microbial components via innate receptors including the C-type lectin receptor Dectin-1, together with the inflammatory environment, programs dendritic cells (DCs) to orchestrate the magnitude and type of adaptive immune responses. The exposure to β-glucan, a known Dectin-1 agonist
Michael Eyres et al.
Cell reports, 40(7), 111230-111230 (2022-08-18)
A defining pathological feature of human lung fibrosis is localized tissue heterogeneity, which challenges the interpretation of transcriptomic studies that typically lose spatial information. Here we investigate spatial gene expression in diagnostic tissue using digital profiling technology. We identify distinct
S J Chapman et al.
Genes and immunity, 11(4), 319-325 (2009-10-03)
The proinflammatory transcription factor nuclear factor-kappaB (NF-kappaB) has a central role in host defence against pneumococcal disease. Both rare mutations and common polymorphisms in the NFKBIA gene encoding the NF-kappaB inhibitor, IkappaB-alpha, associate with susceptibility to bacterial disease, but the
Hendrik Nogai et al.
Blood, 122(13), 2242-2250 (2013-07-23)
Constitutive activation of the nuclear factor-κ B (NF-κB) pathway is a hallmark of the activated B-cell-like (ABC) subtype of diffuse large B-cell lymphoma (DLBCL). Recurrent mutations of NF-κB regulators that cause constitutive activity of this oncogenic pathway have been identified.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service