Skip to Content
Merck
All Photos(5)

Key Documents

HPA007842

Sigma-Aldrich

Anti-CLEC4A antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-C-type lectin DDB27 antibody produced in rabbit, Anti-C-type lectin domain family 4 member A antibody produced in rabbit, Anti-C-type lectin superfamily member 6 antibody produced in rabbit, Anti-Dendritic cell immunoreceptor antibody produced in rabbit, Anti-Lectin-like immunoreceptor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

KNWKSFSSNCYFISTESASWQDSEKDCARMEAHLLVINTQEEQDFIFQNLQEESAYFVGLSDPEGQRHWQWVDQTPYNESSTFWHPREPSDPNERCVVLNFRKSPKRWGWNDVNCLGPQRSVCEMMKIHL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CLEC4A(50856)

Immunogen

C-type lectin domain family 4 member A recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CLEC4A (C-type lectin domain family 4, member A) gene encodes a protein belonging to the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily, the members of which share a common protein fold and are involved in several cellular processes such as cell adhesion, cell-cell signaling, glycoprotein turnover, and immune response. The encoded protein is a type II transmembrane protein expressed on leukocytes and neutrophils. It is involved in the regulation of inflammation in neutrophils. It is found to play a role in the repression of cell activation.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70033

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debleena Pain et al.
The Journal of biological chemistry, 280(8), 6265-6268 (2005-01-11)
Oct4, Nanog, and Stella are transcription factors specifically expressed in embryonic stem (ES) cells and germ lineage cells that impart critical functions in the maintenance of pluripotency. Here, we report the excessive frequency and apparent selectivity of retrotransposition of ES
Manon Richard et al.
Molecular immunology, 43(10), 1716-1721 (2005-12-20)
Proteins that bear immunoreceptor tyrosine based inhibitory motifs (ITIM) are believed to participate in the repression of cell activation via phosphatases such as SHP-1, SHP-2 and/or SHIP-1. CLECSF6, also called DCIR, is a transmembrane protein expressed on leukocytes and predominantly
Liang Chen et al.
JCI insight, 9(12) (2024-05-23)
DC inhibitory receptor (DCIR) is a C-type lectin receptor selectively expressed on myeloid cells, including monocytes, macrophages, DCs, and neutrophils. Its role in immune regulation has been implicated in murine models and human genome-wide association studies, suggesting defective DCIR function
Manon Richard et al.
Biochemical and biophysical research communications, 310(3), 767-773 (2003-10-11)
The recently discovered CLECSF6 protein displays the features of a receptor involved in the down-modulation of leukocyte activation. Although CLECSF6 has been the focus of the interest of many researchers lately, a Western blot characterization of the protein is still
Manon Richard et al.
Journal of leukocyte biology, 71(5), 871-880 (2002-05-08)
In our study of the modulation of the expression of inflammation-related genes in neutrophils, we have found a gene called CLECSF6 (C-type lectin superfamily 6). CLECSF6 expresses two mRNA species at low levels in resting neutrophils. Here, we describe for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service