Skip to Content
Merck
All Photos(4)

Key Documents

HPA007450

Sigma-Aldrich

Anti-SPOCK1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MGC10235, Anti-SPOCK, Anti-TIC1, Anti-sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1, Anti-testican-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

FKDGKLSNNEWCYCFQKPGGLPCQNEMNRIQKLSKGKSLLGAFIPRCNEEGYYKATQCHGSTGQCWCVDKYGNELAGSRKQGAVSCEEEQETSGDFGSGG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SPOCK1(6695)

Immunogen

sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-SPOCK1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

SPOCK1 (sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 1) gene encodes a matricellular glycoprotein that belongs to a family of novel Ca2+-binding proteoglycans. The members of this family contain similar N-terminus, follistatin-like domain, and C-terminus. SPOCK1 is also called as testican-1 and is found to function in cell-cycle regulation, apoptosis, DNA repair, and metastasis. It is a key player in the recurrence of prostate cancer, invasion of glioblastoma, and progression of hepatocellular carcinoma. It activates PI3K/AKT pathway, which in turn, inhibits apoptosis and facilitates proliferation and metastasis of gallbladder cancer cells. It serves as a potential cancer prognostic marker. Defects in this gene may cause developmental delay and microcephaly. This gene may function in neurogenesis. It serves as a metastasis related biomarker in lung carcinoma and may be a potential therapeutic target.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70160

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radhika Dhamija et al.
European journal of medical genetics, 57(4), 181-184 (2014-03-04)
Whole exome sequencing made it possible to identify novel de novo mutations in genes that might be linked to human syndromes (genotype first analysis). We describe a female patient with a novel de novo SPOCK1 variant, which has not been
Liyun Miao et al.
Biochemical and biophysical research communications, 440(4), 792-797 (2013-10-19)
Lung cancer is the leading cause of cancer related death worldwide and the prognosis is still poor with 5-year survival of approximately 15%. Metastasis is the leading cause of death by cancer. Recent researches have demonstrated that epithelial-to-mesenchymal transition (EMT)
Helene Damhofer et al.
Molecular oncology, 7(6), 1031-1042 (2013-09-04)
Pancreatic ductal adenocarcinoma (PDAC) remains one of the most lethal malignancies. It is typically detected at an advanced stage, at which the therapeutic options are very limited. One remarkable feature of PDAC that contributes to its resilience to treatment is
Yung-Wei Lin et al.
Cells, 12(3) (2023-02-12)
Sparc/osteonectin, cwcv, and kazal-like domains proteoglycan 1 (SPOCK1) has been reported to play an oncogenic role in certain cancer types; however, the role of SPOCK1 in the progression of clear cell renal cell carcinoma (ccRCC) remains elusive. Here, higher SPOCK1
Yi-Jun Shu et al.
Molecular cancer, 14, 12-12 (2015-01-28)
Gallbladder cancer (GBC) is a leading cause of cancer-related death worldwide, and its prognosis remains poor, with 5-year survival of approximately 5%. In this study, we analyzed the involvement of a novel proteoglycan, Sparc/osteonectin, cwcv, and kazal-like domains proteoglycan 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service