Skip to Content
Merck
All Photos(5)

Key Documents

HPA007269

Sigma-Aldrich

Anti-SCTR antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-SCT-R, Anti-Secretin receptor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

CDVLQVLWEEQDQCLQELSREQTGDLGTEQPVPGCEGMWDNISCWPSSVPGRMVEVECPRFLRMLTSRNGSLFRNCTQDGWSETFPRPNLACGVNVNDSSNEKRHSYLLKL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SCTR(6344)

Immunogen

Secretin receptor Precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-SCTR antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SCTR (Secretin receptor) is a 50kDa, G protein-coupled receptor (GPCR) associated with the stimulatory cAMP response. It is expressed in the kidney medulla cells. It regulates the ductular bicarbonate, potassium ion from pancreas and enzyme secretion from the pancreatic acinar cells. In the CNS (central nervous system), it facilitates the tyrosine hydroxylase activity that further affects cardiac and renal function.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70127

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anja Klussmeier et al.
Biomedicines, 10(3) (2022-03-26)
Secretin was originally discovered as a gastrointestinal peptide that stimulates fluid secretion from the pancreas and liver and delays gastric emptying. In disease, a secretin receptor (SCTR) was found to occur as a splice variant in gastrinoma and pancreatic adenocarcinoma.
Giuseppe Procino et al.
Kidney international, 86(1), 127-138 (2014-02-14)
X-linked nephrogenic diabetes insipidus (X-NDI) is a disease caused by inactivating mutations of the vasopressin (AVP) type 2 receptor (V2R) gene. Loss of V2R function prevents plasma membrane expression of the AQP2 water channel in the kidney collecting duct cells
B K Chow
Biochemical and biophysical research communications, 212(1), 204-211 (1995-07-06)
Secretin is a gastrointestinal hormone responsible for the regulation of bicarbonate, potassium ion and enzyme secretion from the pancreas. A cDNA encoding the human secretin receptor was isolated from a human pancreatic adenocarcinoma cell-line cDNA library using polymerase chain reaction
D R Patel et al.
Molecular pharmacology, 47(3), 467-473 (1995-03-01)
Secretin is a 27-amino acid neuroendocrine peptide that stimulates fluid and electrolyte secretion in the gastrointestinal tract, activates tyrosine hydroxylase activity in the central nervous system, and affects cardiac and renal function. Specific receptors for secretin have been previously characterized
Combination of secretin and fluvastatin ameliorates the polyuria associated with X-linked nephrogenic diabetes insipidus in mice.
Procino G, Milano S, Carmosino M, et al.
Kidney International, 86(1), 127-138 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service