Skip to Content
Merck
All Photos(4)

Key Documents

HPA007043

Sigma-Aldrich

Anti-HK1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Brain form hexokinase, Anti-HK I, Anti-Hexokinase type I, Anti-Hexokinase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VSGMYLGELVRLILVKMAKEGLLFEGRITPELLTRGKFNTSDVSAIEKNKEGLHNAKEILTRLGVEPSDDDCVSVQHVCTIVSFRSANLVAATLGAILNRLRDNKGTPRLRTTVGVDGSLYKTHPQYSRRFHKTLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HK1(3098)

Immunogen

Hexokinase-1 recombinant protein epitope signature tag (PrEST)

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Biochem/physiol Actions

HK1 (Hexokinase 1) encodes a hexokinase protein consisting of a porin-binding domain at the N-terminal end. It is majorly involved in the glucose metabolic pathway. It is expressed in the photoreceptor inner section, outer plexiform layer, inner nuclear layer, inner plexiform layer, and ganglion cell layer of the human retina. It provides the energy for maintaining the metabolic activity of retina. In the mitochondria, HK1 functions as a pro-survival factor to inhibit TNF-induced apoptotic signals. A missense mutation in hexokinase 1 (HK1) causes an autosomal dominant disorder, retinitis pigmentosa. It has been reported that porin-binding domain of HK1 actively participates in the mitochondrial integrity.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71393

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carsten H Nielsen et al.
Oncotarget, 6(35), 37486-37499 (2015-10-16)
Overexpression of the human epidermal growth factor receptor (HER) family and their ligands plays an important role in many cancers. Targeting multiple members of the HER family simultaneously may increase the therapeutic efficacy. Here, we report the ability to image
Feng Wang et al.
Investigative ophthalmology & visual science, 55(11), 7159-7164 (2014-10-16)
Retinitis pigmentosa (RP) is a genetically heterogeneous disease with over 60 causative genes known to date. Nevertheless, approximately 40% of RP cases remain genetically unsolved, suggesting that many novel disease-causing genes are yet to be identified. In this study, we
Lori S Sullivan et al.
Investigative ophthalmology & visual science, 55(11), 7147-7158 (2014-09-06)
To identify the cause of retinitis pigmentosa (RP) in UTAD003, a large, six-generation Louisiana family with autosomal dominant retinitis pigmentosa (adRP). A series of strategies, including candidate gene screening, linkage exclusion, genome-wide linkage mapping, and whole-exome next-generation sequencing, was used
Koko Murakami et al.
Acta haematologica, 108(4), 204-209 (2002-11-15)
Red blood cells (RBCs) express two hexokinase (HK) isoforms, HK-I and HK-R. Both isozymes are generated from the HK-I gene by use of an alternate promoter. Gene structure and exon-intron organization of the HK-I gene have been elucidated from a
Anja Schindler et al.
Cellular signalling, 25(12), 2685-2692 (2013-09-11)
To coordinate a meaningful response to infection or tissue damage, Tumor Necrosis Factor (TNF) triggers a spectrum of reactions in target cells that includes cell activation, differentiation, proliferation and death. Deregulated TNF signaling can lead to tissue damage and organ

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service