Skip to Content
Merck
All Photos(5)

Documents

HPA005575

Sigma-Aldrich

Anti-TFPI antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-EPI antibody produced in rabbit, Anti-Extrinsic pathway inhibitor antibody produced in rabbit, Anti-LACI antibody produced in rabbit, Anti-Lipoprotein-associated coagulation inhibitor antibody produced in rabbit, Anti-Tissue factor pathway inhibitor precursor antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

QNRFESLEECKKMCTRDNANRIIKTTLQQEKPDFCFLEEDPGICRGYITRYFYNNQTKQCERFKYGGCLGNMNNFETLEECKNICEDGPNGFQVDNYGTQLNAVNNSLTPQSTKVPSLFEFHGPSWCLTPADRGLCRA

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFPI(7035)

General description

Tissue factor pathway inhibitor (TFPI) is a serine proteinase inhibitor. The gene is mapped to human chromosome 2q32. The encoded protein contains three repetitive tandem kunitz-type domains. Domain d1 is involved in binding to the VIIa/TP complex and domain d2 participates in the inhibition of factor Xa. The function of the third domain remains to be characterized. The protein is synthesized in the vascular endothelium.

Immunogen

Tissue factor pathway inhibitor precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-TFPI antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Tissue factor pathway inhibitor (TFPI), also referred to as EPI, down regulates tissue factor initiated blood coagulation. It has a nonspecific association with cell surface glycosaminoglycans and binding to glycosyl phosphatidylinositol-anchored proteins. It also binds specifically and saturably to thrombospondin-1 (TSP-1). TFPI may be involved in inhibiting factor X and VIIa, which are blood clotting factors, thus possessing an antithrombotic action and also the ability to associate with lipoproteins in plasma.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST83059

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tissue Factor Pathway Inhibitor (TFPI): Biochemistry, Genetics, and Implications for Thrombophilia.
Brinkmann T
LaboratoriumsMedizin/Journal of Laboratory Medicine, 25, 269-276 null
Refined regional assignment of the human tissue factor pathway inhibitor (TFPI) gene to chromosome band 2q32 by non-isotopic in situ hybridization.
C P Van der Logt et al.
Human genetics, 89(5), 577-578 (1992-07-01)
Bashir A Lwaleed et al.
The Journal of pathology, 208(3), 327-339 (2005-11-02)
Tissue factor (TF)-initiated coagulation plays a significant role in the pathophysiology of many diseases, including cancer and inflammation. Tissue factor pathway inhibitor (TFPI) is a plasma Kunitz-type serine protease inhibitor, which modulates initiations of coagulation induced by TF. In a
A E Mast et al.
The Journal of biological chemistry, 275(41), 31715-31721 (2000-08-03)
Tissue factor pathway inhibitor (TFPI) is a Kunitz-type serine proteinase inhibitor that down-regulates tissue factor-initiated blood coagulation. The most biologically active pool of TFPI is associated with the vascular endothelium, however, the biochemical mechanisms responsible for its cellular binding are
Dae-Jin Kwon et al.
Transgenic research, 26(1), 153-163 (2016-08-25)
Recent progress in genetic manipulation of pigs designated for xenotransplantation ha6s shown considerable promise on xenograft survival in primates. However, genetic modification of multiple genes in donor pigs by knock-out and knock-in technologies, aiming to enhance immunological tolerance against transplanted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service