Skip to Content
Merck
All Photos(6)

Documents

HPA005157

Sigma-Aldrich

Anti-TYK2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Non-receptor tyrosine-protein kinase TYK2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

RFLRDFQPGRLSQQMVMVKYLATLERLAPRFGTERVPVCHLRLLAQAEGEPCYIRDSGVAPTDPGPESAAGPPTHEVLVTGTGGIQWWPVEEEVNKEEGSSGSSGRNPQASLFGKKAKAHKAFGQPADRPREPLWAYFCDFRDITHVV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TYK2(7297)

General description

Tyrosine kinase 2 (TYK2) belongs to the Janus Kinase (Jak) family and is a non-receptor tyrosine kinase. TYK2 gene maps to chromosome 19p13.2 and is expressed in non-hematopoietic as well as hematopoietic cell lines. TYK2 gene has high variations in human population, with more than 100 non-synonymous TYK2 gene variants present.

Immunogen

Non-receptor tyrosine-protein kinase TYK2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

TYK2 (Tyrosine kinase 2) plays an important role in cell signaling by multiple cytokines via the JAK-STAT pathway. Tyk2 interacts with type I IFN, p40-sharing IL-12 and IL-23, IL-6, and IL-10 family receptors and helps in immunomodulation. On binding, TYK2 is phosphorylated and co-activated with JAK1 and/or JAK2. It has been suggested that TYK2 is required for the surface expression of human IL-10R as well as human type I IFNR. TYK2 is also involved in the differentiation of immune cells, such as B cells, T helper cells and natural killer cells. Abnormal TYK2 expression is implicated in the etiology of several auto-immune diseases such as multiple sclerosis, rheumatoid arthritis and systemic lupus erythematosus. Studies show that TYK2 rs34536443 polymorphism is associated with endometriosis and related infertility.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST78871

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoshiyuki Minegishi et al.
Immunity, 25(5), 745-755 (2006-11-08)
Tyrosine kinase 2 (Tyk2) is a nonreceptor tyrosine kinase that belongs to the Janus kinase (Jak) family. Here we identified a homozygous Tyk2 mutation in a patient who had been clinically diagnosed with hyper-IgE syndrome. This patient showed unusual susceptibility
Yan Liang et al.
Expert opinion on therapeutic targets, 18(5), 571-580 (2014-03-25)
Tyrosine kinase 2 (Tyk2) is a Janus kinase family member that is crucial for signaling transduction in response to a wide variety of cytokines, including type I IFNs, IL-6, IL-10, IL-12 and IL-23. An appropriate expression of Tyk2-mediated signaling might
Sue J Sohn et al.
Journal of immunology (Baltimore, Md. : 1950), 191(5), 2205-2216 (2013-07-31)
TYK2 is a JAK family protein tyrosine kinase activated in response to multiple cytokines, including type I IFNs, IL-6, IL-10, IL-12, and IL-23. Extensive studies of mice that lack TYK2 expression indicate that the IFN-α, IL-12, and IL-23 pathways, but
Zhi Li et al.
Journal of immunology (Baltimore, Md. : 1950), 190(5), 2335-2344 (2013-01-30)
Tyk2 belongs to the Janus protein tyrosine kinase family and is involved in signaling of immunoregulatory cytokines (type I and III IFNs, IL-6, IL-10, and IL-12 families) via its interaction with shared receptor subunits. Depending on the receptor complex, Tyk2
Carla Peluso et al.
Human immunology, 74(1), 93-97 (2012-09-25)
Tyrosine kinase 2 gene (TYK2) is part of the janus kinase (JAK) that binds to the type I interferon-α receptor (IFNAR) on the cell surface of IFN-producing cells, and have crucial importance in the etiology of autoimmune and inflammatory diseases.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service