Skip to Content
Merck
All Photos(9)

Key Documents

HPA004428

Sigma-Aldrich

Anti-ACAT1 antibody produced in rabbit

enhanced validation

Ab1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

ACAT, THIL

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

mouse, human, rat

enhanced validation

RNAi knockdown
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VSATRTPIGSFLGSLSLLPATKLGSIAIQGAIEKAGIPKEEVKEAYMGNVLQGGEGQAPTRQAVLGAGLPISTPCTTINKVCASGMKAIMMASQSLMCGHQDVMVAGGMESMSNVPYVMNRGSTPYGGVKLEDLIVKDGLTD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACAT1(38)

Looking for similar products? Visit Product Comparison Guide

General description

The 56kDa protein ACAT1 (acetyl-CoA acetyltransferase 1) is majorly involved in the cellular cholesterol homeostasis. It belongs to the membrane-bound O-acyltransferase family, presents in various cell types. In mammals, ACAT1 exists in two isoenzyme form: ACAT1 and ACAT2. It is composed of nine transmembrane domains (TMDs), with two active sites. One active site His460, is located within TMD7 and the second active site Asn421, is located within the fourth large cytoplasmic loop.

Immunogen

Acetyl-CoA acetyltransferase, mitochondrial precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-ACAT1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

ACAT1 (acetyl-CoA acetyltransferase 1) is highly involved in the ketogenic pathway. It accelerates the conversion of cholesterol to cholesteryl esters. The elevated level of the protein expression has been observed in the prostate cancer. Thus, it acts as a potential prognostic marker in prostate cancer.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86794

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aicha Saadane et al.
The Journal of biological chemistry, 291(39), 20427-20439 (2016-08-16)
The process of vision is impossible without the photoreceptor cells, which have a unique structure and specific maintenance of cholesterol. Herein we report on the previously unrecognized cholesterol-related pathway in the retina discovered during follow-up characterizations of Cyp27a1(-/-)Cyp46a1(-/-) mice. These
Svenia Schnyder et al.
American journal of physiology. Endocrinology and metabolism, 312(5), E437-E446 (2017-02-23)
Low-carbohydrate/high-fat (LCHF) diets are increasingly popular dietary interventions for body weight control and as treatment for different pathological conditions. However, the mechanisms of action are still poorly understood, in particular, in long-term administration. Besides liver, brain, and heart, skeletal muscle
Kristoffer Svensson et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(5), 1976-1986 (2016-02-07)
Ketone bodies (KBs) are crucial energy substrates during states of low carbohydrate availability. However, an aberrant regulation of KB homeostasis can lead to complications such as diabetic ketoacidosis. Exercise and diabetes affect systemic KB homeostasis, but the regulation of KB
Punit Saraon et al.
The Prostate, 74(4), 372-380 (2013-12-07)
Prostate cancer is the second leading cause of cancer-related death among men in North America. While a majority of prostate cancer cases remain indolent, subsets of patients develop aggressive cancers, which may lead to death. The current methods of detection
Guang-Jing Hu et al.
Cell research, 23(8), 1007-1024 (2013-07-10)
Trans-splicing, a process involving the cleavage and joining of two separate transcripts, can expand the transcriptome and proteome in eukaryotes. Chimeric RNAs generated by trans-splicing are increasingly described in literatures. The widespread presence of antibiotic resistance genes in natural environments

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service