Skip to Content
Merck
All Photos(1)

Documents

HPA003506

Sigma-Aldrich

Anti-HMGB2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-HMG-2 antibody produced in rabbit, Anti-High mobility group protein 2 antibody produced in rabbit, Anti-High mobility group protein B2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50- 1:200

immunogen sequence

KSKFEDMAKSDKARYDREMKNYVPPKGDKKGKKKDPNAPKRPPSAFFLFCSEHRPKIKSEHPGLSIGDTAKKLGEMWSEQSAKDKQPYEQKAAKLKEKYE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HMGB2(3148)

General description

High-mobility group protein B2, also known as high-mobility group protein 2 (HMG-2), is a protein encoded by the HMGB2 gene in humans. The protein belongs to a family of chromatin proteins made up of two basic DNA binding domains, HMG box A and B, and a C-terminal acidic tail. It consists of highly conserved sequence, but different expression pattern.

Immunogen

High mobility group protein B2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

High-mobility group protein B2 (HMGB2) functions as a significant prognostic factor and may play a crucial role in cell invasion. The gene acts as a novel prognostic marker and an attractive therapeutic target for glioblastoma multiforme (GBM). The protein helps in altering DNA elasticity while facilitating transcription, replication and DNA repair. It plays a significant role in tumor development and during prognosis of hepatocellular carcinoma (HCC). It may also be associated with the antiapoptotic pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86405

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alok Tripathi et al.
Bioinformation, 15(7), 467-473 (2019-09-06)
Recently, High Mobility Group Box1 (HMGB1) protein has been reported as an inflammatory cytokine present in all nucleated cells with crucial role in the genesis and promotion of cancer. No HMGB1 protein mice model and its active site details are
Zhe Bao Wu et al.
Neuro-oncology, 15(9), 1264-1275 (2013-07-06)
The expression profile of high-mobility group box 2 (HMGB2) in patients with glioblastoma multiforme (GBM) and its clinical signature with underlying mechanisms were not fully explored. HMGB2 protein levels were measured in 51 GBM patients by immunohistochemical studies. To clarify
Nazia Syed et al.
Proteomics, 15(2-3), 383-393 (2014-10-21)
Dysregulation of protein expression is associated with most diseases including cancer. MS-based proteomic analysis is widely employed as a tool to study protein dysregulation in cancers. Proteins that are differentially expressed in head and neck squamous cell carcinoma (HNSCC) cell
Tobias Pusterla et al.
Autoimmunity, 42(4), 308-310 (2009-10-09)
High mobility group B box (HMGB) proteins are a family of chromatin proteins made up of two basic DNA binding domains, HMG box A and B, and a C-terminal acidic tail. HMGB have a highly conserved sequence, but different expression
Jung-Hee Kwon et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 16(22), 5511-5521 (2010-09-21)
We investigated the expression of high-mobility group box 2 (HMGB2) in patients with hepatocellular carcinoma (HCC) and its clinical effects with underlying mechanisms. HMGB2 mRNA levels were measured in 334 HCC patients by real-time reverse transcription-PCR and HMGB2 protein levels

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service