Skip to Content
Merck
All Photos(2)

Documents

HPA002320

Sigma-Aldrich

Anti-LMAN1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-ER-Golgi intermediate compartment 53 kDa protein antibody produced in rabbit, Anti-ERGIC-53 protein precursor antibody produced in rabbit, Anti-Gp58 antibody produced in rabbit, Anti-Intracellular mannose-specific lectin MR60 antibody produced in rabbit, Anti-Lectin, mannose-binding 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

IGNNGQIHYDHQNDGASQALASCQRDFRNKPYPVRAKITYYQNTLTVMINNGFTPDKNDYEFCAKVENMIIPAQGHFGISAATGGLADDHDVLSFLTFQLTEPGKEP

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LMAN1(3998)

General description

Lectin mannose-binding 1 (LMAN1) is a mannose-specific, calcium-dependent lectin-like transport receptor protein.

Immunogen

ERGIC-53 protein precursor recombinant protein epitope signature tag (PrEST)

Application

Anti-LMAN1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

LMAN1 mediates intracellular transport of bidirectional glycoproteins from the endoplasmic reticulum to Golgi body. It interacts with cathepsin-Z-related proteins for the intracellular trafficking of the lysosomal enzyme cathepsin C and for efficient secretion of coagulation factors V and VIII. The efficiency of the gene in terms of expression level as well as recycling pathway is modulated by heat shock during cell stress. Heat shock enhances the translation of the LMAN1 mRNA followed by accumulation of LMAN1. Mutated LMAN1 leads to an autosomal recessive bleeding disorder i.e. combined deficiency of factor V and factor VIII (F5F8D). It has also been reported that abnormalities of LMAN1 potentially contribute to microsatellite instability (MSI)-H carcinogenesis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86408

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carmen Spatuzza et al.
The Journal of biological chemistry, 279(41), 42535-42544 (2004-08-05)
ERGIC-53 is a lectin-like transport receptor protein, which recirculates between the ER and the Golgi complex and is required for the intracellular transport of a restricted number of glycoproteins. We show in this article that ERGIC-53 accumulates during the heat
Lili-Naz Hazrati et al.
Neurobiology of aging, 33(12), 2949-2949 (2012-07-24)
CR1 is a novel Alzheimer's disease (AD) gene identified by genome-wide association studies (GWAS). Recently, we showed that AD risk could be explained by an 18-kilobase insertion responsible for the complement component (3b/4b) receptor 1 (CR1)-S isoform. We investigated the
Nina Roeckel et al.
Cancer research, 69(1), 292-299 (2009-01-02)
Glycosyl epitopes have been identified as tumor-specific markers in colorectal tumors and various lines of evidence indicate the significance of altered synthesis, transport, and secretion of glycoproteins in tumorigenesis. However, aberrant glycosylation has been largely ignored in microsatellite unstable (MSI-H)
Bin Zhang et al.
Nature genetics, 34(2), 220-225 (2003-04-30)
Mutations in LMAN1 (also called ERGIC-53) result in combined deficiency of factor V and factor VIII (F5F8D), an autosomal recessive bleeding disorder characterized by coordinate reduction of both clotting proteins. LMAN1 is a mannose-binding type 1 transmembrane protein localized to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service